PloS one
Renal primordia activate kidney regenerative events in a rat model of progressive renal disease.
PMID 25811887
Abstract
New intervention tools for severely damaged kidneys are in great demand to provide patients with a valid alternative to whole organ replacement. For repairing or replacing injured tissues, emerging approaches focus on using stem and progenitor cells. Embryonic kidneys represent an interesting option because, when transplanted to sites such as the renal capsule of healthy animals, they originate new renal structures. Here, we studied whether metanephroi possess developmental capacity when transplanted under the kidney capsule of MWF male rats, a model of spontaneous nephropathy. We found that six weeks post-transplantation, renal primordia developed glomeruli and tubuli able to filter blood and to produce urine in cyst-like structures. Newly developed metanephroi were able to initiate a regenerative-like process in host renal tissues adjacent to the graft in MWF male rats as indicated by an increase in cell proliferation and vascular density, accompanied by mRNA and protein upregulation of VEGF, FGF2, HGF, IGF-1 and Pax-2. The expression of SMP30 and NCAM was induced in tubular cells. Oxidative stress and apoptosis markedly decreased. Our study shows that embryonic kidneys generate functional nephrons when transplanted into animals with severe renal disease and at the same time activate events at least partly mimicking those observed in kidney tissues during renal regeneration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H7157 Anti-Hepatocyte Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
H0652 Anti-Hepatocyte Growth Factor antibody produced in goat, IgG fraction of antiserum, lyophilized powder
H0540 Anti-Hepatocyte Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
H9786 Anti-Hepatocyte Growth Factor Receptor (c-Met) antibody produced in goat, affinity isolated antibody, lyophilized powder
H9911 Anti-Hepatocyte Growth Factor Receptor antibody produced in goat, lyophilized powder, affinity isolated antibody
SAB2101033
Anti-HGF (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101034
Anti-HGF (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2105571
Anti-HGF antibody produced in rabbit, affinity isolated antibody
HPA040360
Anti-HGF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA044088
Anti-HGF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410624
Anti-HGF antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA005980
Anti-LRP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300599
Anti-MET (Ab-1003) antibody produced in rabbit, affinity isolated antibody
SAB4300479
Anti-MET (Ab-1234) antibody produced in rabbit, affinity isolated antibody
SAB4300487
Anti-MET (Ab-1349) antibody produced in rabbit, affinity isolated antibody
SAB1406117
Anti-MET antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4501870
Anti-MET antibody produced in rabbit, affinity isolated antibody
SAB4501868
Anti-MET, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501869
Anti-MET, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2700234
Anti-Ncam1 (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2501672
Anti-NCAM1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4300660
Anti-NCAM1 antibody produced in rabbit, affinity isolated antibody
AV100859
Anti-PAX2 antibody produced in rabbit, IgG fraction of antiserum
SAB2101729
Anti-PAX2 antibody produced in rabbit, affinity isolated antibody
HPA047704
Anti-PAX2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106223
Anti-PAX2 antibody produced in rabbit, affinity isolated antibody
SAB1410876
Anti-PAX2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
V6627 Anti-Vascular Endothelial Growth Factor antibody produced in goat, IgG fraction of antiserum, lyophilized powder
V1253 Anti-Vascular Endothelial Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB2102716
Anti-WT1 antibody produced in rabbit, affinity isolated antibody