EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Biodegradation of fenoxaprop-P-ethyl (FE) by Acinetobacter sp. strain DL-2 and cloning of FE hydrolase gene afeH.
PMID 25812814
Abstract
Fenoxaprop-P-ethyl (FE) is widely used as a post-emergence aryloxyphenoxy propionate (AOPP) herbicide in agriculture. An efficient FE-degrading strain DL-2 was isolated from the enrichment culture and identified as Acinetobacter sp. and the metabolite fenoxaprop acid (FA) was identified by HPLC/MS analysis. The strain DL-2 could also degrade a wide range of other AOPP herbicides. A novel FE hydrolase esterase gene afeH was cloned from strain DL-2 and functionally expressed in Escherichia coli BL21(DE3). The specific activities of recombinant AfeH was 216.39 U mg(-1) for FE with Km and Vmax values of 0.82 μM and 7.94 μmol min(-1) mg(-1). AfeH could also hydrolyze various AOPP herbicides, p-nitrophenyl esters and triglycerides. The optimal pH and temperature for recombinant AfeH were 9.0 and 50°C, respectively; the enzyme was activated by Co(2+) and inhibited by Ca(2+), Zn(2+), Ba(2+). AfeH was inhibited strongly by phenylmethylsulfonyl and SDS and weakly by dimethyl sulfoxide.