EMAIL THIS PAGE TO A FRIEND
Investigative radiology
Feasibility of Selective Catheter-Directed Coronary Computed Tomography Angiography Using Ultralow-Dose Intracoronary Contrast Injection in a Swine Model.
PMID 25816215
Abstract
Selective catheter-directed intracoronary contrast injected coronary computed tomography angiography (selective CCTA) has recently been introduced for on-site evaluation of coronary artery disease during coronary artery catheterization. In this study, we aimed to develop a feasible protocol for selective CCTA using ultralow-dose contrast medium as compared with conventional intravenous CCTA (IV CCTA). A novel combined system incorporating coronary angiography and a 320-detector row computed tomographic scanner was used to study 4 swine (35-40 kg) under animal institutional review board approval. A selective CCTA scan was simultaneously performed with an injection of 13.13 mgI/mL of modulated contrast medium at multiple different injection rates including 2, 3, and 4 mL/s and different total injection volumes of either 20 or 30 mL. Intravenous CCTA was performed with 60 mL of contrast medium, followed by 30 mL of saline chaser at 5 mL/s. Coronary mean and peak intensity, transluminal attenuation gradient, as well as 3-dimensional maximum intensity projections were obtained. Attenuation values (mean ± standard error, in Hounsfield units [HUs]) of selective CCTA for the left anterior descending (LAD) and right coronary artery (RCA) using the various combinations of injection rates and total injection volumes were as follows: 20 mL at 2 mL/s (LAD, 270.3 ± 20.4 HU; RCA, 322.6 ± 7.4 HU), 20 mL at 3 mL/s (LAD, 262.9 ± 20.4 HU; RCA, 264.7 ± 7.4 HU), 30 mL at 3 mL/s (LAD, 276.8 ± 20.4 HU; RCA, 274.0 ± 7.4 HU), 20 mL at 4 mL/s (LAD, 268.0 ± 20.4 HU; RCA, 277.7 ± 7.4 HU), and 30 mL at 4 mL/s (LAD, 251.3 ± 20.4 HU; RCA, 334.7 ± 7.4 HU). The representative protocol of the selective CCTA studies produced results within the optimal enhancement range (approximately 250-350 HU) for all segments, and comparison of transluminal attenuation gradient data with selective CCTA and IV CCTA studies demonstrated that the former method was more homogenous (-1.5245 and -1.7558 for LAD as well as 0.0459 and 0.0799 for RCA, respectively). Notably, the volume of iodine contrast medium used for selective CCTA was reported to be 1.09% (0.2 g) of IV CCTA (24 g). The current findings demonstrate the feasibility of selective CCTA using ultralow-dose intracoronary contrast injection. This technique may provide additional means of coronary evaluation in patients who may require strategic planning before a procedure using a combined modality system.