EMAIL THIS PAGE TO A FRIEND
Neurobiology of aging
Visual impairment in an optineurin mouse model of primary open-angle glaucoma.
PMID 25818176
Abstract
Primary open-angle glaucoma (POAG) is characterized by progressive neurodegeneration of retinal ganglion cells (RGCs). Why RGCs degenerate in low-pressure POAG remains poorly understood. To gain mechanistic insights, we developed a novel mouse model based on a mutation in human optineurin associated with hereditary, low-pressure POAG. This mouse improves the design and phenotype of currently available optineurin mice, which showed high global overexpression. Although both 18-month-old optineurin and nontransgenic control mice showed an age-related decrease in healthy axons and RGCs, the expression of mutant optineurin enhanced axonal degeneration and decreased RGC survival. Mouse visual function was determined using visual evoked potentials, which revealed specific visual impairment in contrast sensitivity. The E50K optineurin transgenic mouse described here exhibited clinical features of POAG and may be useful for mechanistic dissection of POAG and therapeutic development.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9979
Anti-Aly antibody, Mouse monoclonal, clone 11G5, purified from hybridoma cell culture
HPA007305
Anti-CALB2 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA007306
Anti-CALB2 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103301
Anti-CALB2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2500188
Anti-Calretinin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
C7479
Anti-Calretinin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701734
Anti-cdk7 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA007932
Anti-CDK7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV03009
Anti-CDK7 antibody produced in rabbit, IgG fraction of antiserum
SAB2104125
Anti-CDK7 antibody produced in rabbit, affinity isolated antibody
SAB4500060
Anti-CDK7 antibody produced in rabbit, affinity isolated antibody
SAB4500061
Anti-CDK7, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300593
Anti-KIT (Ab-936) antibody produced in rabbit, affinity isolated antibody
SAB1412042
ANTI-KIT antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
SAB1412043
ANTI-KIT antibody produced in mouse, clone 5F6, purified immunoglobulin, buffered aqueous solution
SAB1412044
ANTI-KIT antibody produced in mouse, clone 5A11, purified immunoglobulin, buffered aqueous solution
SAB1412045
ANTI-KIT antibody produced in mouse, clone 2C3, purified immunoglobulin, buffered aqueous solution
SAB1412046
ANTI-KIT antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
SAB1412140
ANTI-KIT antibody produced in mouse, clone 4F7, purified immunoglobulin, buffered aqueous solution
SAB1412141
ANTI-KIT antibody produced in mouse, clone 3A8, purified immunoglobulin, buffered aqueous solution
SAB4501647
Anti-KIT, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1411601
Anti-MAP3K14 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1306410
ANTI-NIK (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
N0149 Anti-NIK (NF-κB Inducing Kinase) antibody produced in rabbit, 1 mg/mL, IgG fraction of antiserum, buffered aqueous solution
PRS1129
Anti-NIK antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1306052
ANTI-NIK(C-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4700750
Monoclonal Anti-CD117 antibody produced in mouse, clone 104D2, purified immunoglobulin, buffered aqueous solution
WH0001022M1 Monoclonal Anti-CDK7 antibody produced in mouse, clone 1G5, purified immunoglobulin, buffered aqueous solution
WH0003815M2
Monoclonal Anti-KIT antibody produced in mouse, clone 6F2, purified immunoglobulin, buffered aqueous solution
AMAB90901
Monoclonal Anti-KIT antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1657, purified immunoglobulin, buffered aqueous glycerol solution