EMAIL THIS PAGE TO A FRIEND
Cell reports
Prostate tumorigenesis induced by PTEN deletion involves estrogen receptor β repression.
PMID 25818291
Abstract
The role of ERβ in prostate cancer is unclear, although loss of ERβ is associated with aggressive disease. Given that mice deficient in ERβ do not develop prostate cancer, we hypothesized that ERβ loss occurs as a consequence of tumorigenesis caused by other oncogenic mechanisms and that its loss is necessary for tumorigenesis. In support of this hypothesis, we found that ERβ is targeted for repression in prostate cancer caused by PTEN deletion and that loss of ERβ is important for tumor formation. ERβ transcription is repressed by BMI-1, which is induced by PTEN deletion and important for prostate tumorigenesis. This finding provides a mechanism for how ERβ expression is regulated inxa0prostate cancer. Repression of ERβ contributes to tumorigenesis because it enables HIF-1/VEGF signaling that sustains BMI-1 expression. These data reveal a positive feedback loop that is activated in response to PTEN loss and sustains BMI-1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN1021 CompoZr® Knockout ZFN Kit, human ESR2 (NM_001437)