EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and pharmacology
Naringin attenuates the cytotoxicity of hepatotoxin microcystin-LR by the curious mechanisms to OATP1B1- and OATP1B3-expressing cells.
PMID 25818985
Abstract
Microcystin-LR, which is an inhibitor of serine/threonine protein phosphatase (PP)1 and PP2A, induces liver injury by its selective uptake system into the hepatocyte. It is also thought that microcystin-LR induces reactive oxygen species (ROS). We tried to establish the chemical prevention of microcystin-LR poisoning. We investigated the effect of grapefruit flavanone glycoside naringin on cytotoxicity of microcystin-LR using human hepatocyte uptake transporter OATP1B3-expressing HEK293-OATP1B3 cells. We found cytotoxicity of microcystin-LR was attenuated by naringin in a dose dependent manner. The inhibition magnitude of total cellular serine/threonine protein phosphatase activity induced by microcystin-LR was suppressed by naringin. In addition, uptake of microcystin-LR into HEK293-OATP1B3 cells was inhibited by naringin. Furthermore, microcystin-LR induced phosphorylation of p53 was inhibited by naringin. Regardless of the difference in the exposure pattern of pre-processing and post-processing of naringin, the toxicity of microcystin-LR was comparable. These results suggested that naringin is promising remedy as well as preventive medicine for liver damage with microcystin-LR. In addition, involvement of ROS production after exposure to the sublethal concentrations of microcystin-LR in the onset of cytotoxicity was negligible. Therefore, inhibition of microcystin-LR uptake and the pathway other than ROS production would be involved in the effect of naringin on the attenuation of microcystin-LR toxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1376
Naringin, ≥90%, from citrus fruit
C27H32O14
71162
Naringin, ≥95% (HPLC)
C27H32O14