EMAIL THIS PAGE TO A FRIEND
Experimental neurology
Lesion-induced accumulation of platelets promotes survival of adult neural stem / progenitor cells.
PMID 25819103
Abstract
The presence of neural stem/progenitor cells (NSPCs) in specific areas of the central nervous system (CNS) supports tissue maintenance as well as regeneration. The subependymal zone (SEZ), located at the lateral ventricle's wall, represents a niche for NSPCs and in response to stroke or demyelination becomes activated with progenitors migrating towards the lesion and differentiating into neurons and glia. The mechanisms that underlie this phenomenon remain largely unknown. The vascular niche and in particular blood-derived elements such as platelets, has been shown to contribute to CNS regeneration in different pathological conditions. Indeed, intracerebroventricularly administrated platelet lysate (PL) stimulates angiogenesis, neurogenesis and neuroprotection in the damaged CNS. Here, we explored the presence of platelets in the activated SEZ after a focal demyelinating lesion in the corpus callosum of mice and we studied the effects of PL on proliferating SEZ-derived NSPCs in vitro. We showed that the lesion-induced increase in the size of the SEZ and in the number of proliferating SEZ-resident NSPCs correlates with the accumulation of platelets specifically along the activated SEZ vasculature. Expanding on this finding, we demonstrated that exposure of NSPCs to PL in vitro led to increased numbers of cells by enhanced cell survival and reduced apoptosis without differences in proliferation and in the differentiation potential of NSPCs. Finally, we demonstrate that the accumulation of platelets within the SEZ is spatially correlated with reduced numbers of apoptotic cells when compared to other periventricular areas. In conclusion, our results show that platelet-derived compounds specifically promote SEZ-derived NSPC survival and suggest that platelets might contribute to the enlargement of the pool of SEZ NSPCs that are available for CNS repair in response to injury.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1303049 ANTI-BHLHB5 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
AV32430
Anti-BHLHB5 antibody produced in rabbit, affinity isolated antibody
SAB2106261
Anti-BHLHB5 antibody produced in rabbit, affinity isolated antibody
SAB1412832
Anti-BHLHE22 antibody produced in mouse, purified immunoglobulin, 0.5 mg/mL
SAB1405545
Anti-CACNB3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV34957
Anti-CACNB3 antibody produced in rabbit, IgG fraction of antiserum
SAB2100332
Anti-CACNB3 antibody produced in rabbit, affinity isolated antibody
SAB1411416
Anti-CACNB3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501404 Anti-GABRB3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2100880
Anti-GABRB3 antibody produced in rabbit, affinity isolated antibody
AV32753
Anti-OLIG2 (AB1) antibody produced in rabbit, affinity isolated antibody
AV31464
Anti-OLIG2 (AB2) antibody produced in rabbit, affinity isolated antibody
AV32752
Anti-OLIG2 (AB3) antibody produced in rabbit, affinity isolated antibody
SAB1412293
ANTI-OLIG2 antibody produced in mouse, clone 2A8, purified immunoglobulin, buffered aqueous solution
SAB1412294
ANTI-OLIG2 antibody produced in mouse, clone 2B11, purified immunoglobulin, buffered aqueous solution
SAB5200049
Monoclonal Anti-GABRB3 antibody produced in mouse, clone S87-25, 1 mg/mL, purified immunoglobulin
WH0010215M2
Monoclonal Anti-OLIG2 antibody produced in mouse, clone 3D7, purified immunoglobulin, buffered aqueous solution
SAB1404798
Monoclonal Anti-OLIG2 antibody produced in mouse, clone 3C9, purified immunoglobulin, buffered aqueous solution
SAB5300407
Monoclonal Anti-OLIG2 antibody produced in mouse, clone 1G11, ascites fluid