EMAIL THIS PAGE TO A FRIEND
Oncotarget
Ndrg2 promoter hypermethylation triggered by helicobacter pylori infection correlates with poor patients survival in human gastric carcinoma.
PMID 25823664
Abstract
N-myc downstream regulated gene 2 (Ndrg2) is a candidate suppressor of cancer metastasis. We found that Ndrg2 promoter was frequently hypermethylated in gastric cancer cell lines and in 292 gastric tumor tissues. This resulted in down-regulation of Ndrg2 mRNA and protein. Ndrg2 promoter methylation was associated with H. pylori infection and worse prognosis of gastric cancer patients, which is an independent prognostic factor for the disease-free survival (DFS). We found that H. pylori silenced Ndrg2 by activating the NF-κB pathway and up-regulating DNMT3b, promoting gastric cancer progression. These findings uncover a previously unrecognized role for H. pylori infection in gastric cancer.