EMAIL THIS PAGE TO A FRIEND
Carcinogenesis
A genetic variant in CHRNB3-CHRNA6 increases risk of esophageal squamous cell carcinoma in Chinese populations.
PMID 25823894
Abstract
Nicotinic acetylcholine receptors are important regulators of smoking behavior and tobacco carcinogenesis. We studied the association of the CHRNB3-A6 variant rs13280604 in relation to esophageal squamous cell carcinoma (ESCC) in Chinese populations. Two independent case-control studies were conducted. The first case-control study, consisted of 866 ESCC patients and 1621 healthy controls from Northern China, and the second case-control study consisted of 853 ESCC patients and 860 unrelated controls from Southern China. A logistic regression model was used to evaluate the associations of rs13280604 with cancer risk. We found that Rs13280604 GG/AG genotypes were significantly associated with increased risk for ESCC in both case-control studies from Northern [odds ratio (OR), 1.42, 95% confidence interval (CI), 1.19-1.70, P = 1.1×10(-4)], Southern China (OR, 1.56, 95% CI, 1.26-1.93, P = 5.2×10(-5)), and the combined population of both studies (OR, 1.44, 95% CI, 1.26-1.65, P = 8.7×10(-8)), respectively. Our results suggest that this CHRNB3-A6 variant confers susceptibility to ESCC risk. However, future larger studies are needed to validate our finding.