EMAIL THIS PAGE TO A FRIEND
Nature communications
Global analysis of fungal morphology exposes mechanisms of host cell escape.
PMID 25824284
Abstract
Developmental transitions between single-cell yeast and multicellular filaments underpin virulence of diverse fungal pathogens. For the leading human fungal pathogen Candida albicans, filamentation is thought to be required for immune cell escape via induction of an inflammatory programmed cell death. Here we perform a genome-scale analysis of C. albicans morphogenesis and identify 102 negative morphogenetic regulators and 872 positive regulators, highlighting key roles for ergosterol biosynthesis and N-linked glycosylation. We demonstrate that C. albicans filamentation is not required for escape from host immune cells; instead, macrophage pyroptosis is driven by fungal cell-wall remodelling and exposure of glycosylated proteins in response to the macrophage phagosome. The capacity of killed, previously phagocytized cells to drive macrophage lysis is also observed with the distantly related fungal pathogen Cryptococcus neoformans. This study provides a global view of morphogenetic circuitry governing a key virulence trait, and illuminates a new mechanism by which fungi trigger host cell death.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000005 Amphotericin B, European Pharmacopoeia (EP) Reference Standard
C47H73NO17
A4888
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
C47H73NO17
M9546
D-Mannitol, BioXtra, ≥98% (GC)
C6H14O6
M9647
D-Mannitol, ACS reagent
C6H14O6
M4125
D-Mannitol, ≥98%
C6H14O6
M1902
D-Mannitol, plant cell culture tested
C6H14O6
M8429
D-Mannitol, meets EP, FCC, USP testing specifications
C6H14O6
63559
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
C6H14O6
63560
D-Mannitol, ACS reagent, for microbiology, ≥99.0%
C6H14O6
17311
D-Mannitol, tested according to Ph.Eur.
C6H14O6
78513
D-Mannitol, ≥99.9999% (metals basis), for boron determination
C6H14O6
F8929
Fluconazole, ≥98% (HPLC), powder
C13H12F2N6O
PHR1160
Fluconazole, Pharmaceutical Secondary Standard; Certified Reference Material
C13H12F2N6O
1271700
Fluconazole, United States Pharmacopeia (USP) Reference Standard
C13H12F2N6O
Y0000557
Fluconazole, European Pharmacopoeia (EP) Reference Standard
C13H12F2N6O
Y0000558
Fluconazole for peak identification, European Pharmacopoeia (EP) Reference Standard
C13H12F2N6O
PHR1007
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
C6H14O6
1375105
Mannitol, United States Pharmacopeia (USP) Reference Standard
C6H14O6
M0200000
Mannitol, European Pharmacopoeia (EP) Reference Standard
C6H14O6
P2222
Potassium phosphate dibasic, meets USP testing specifications
HK2O4P
450200
Potassium phosphate dibasic, 99.95% trace metals basis
HK2O4P
P8281
Potassium phosphate dibasic, reagent grade, ≥98.0%
HK2O4P
60353
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
HK2O4P
60356
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
HK2O4P
P8584
Potassium phosphate dibasic solution, 1.0 M
HK2O4P
T8826
Terbinafine hydrochloride, ≥98%
C21H25N · HCl
T7765
Tunicamycin from Streptomyces sp.