EMAIL THIS PAGE TO A FRIEND
European journal of immunology
Inhibition of NKp30- and 2B4-mediated NK cell activation by evolutionary different human and bovine CEACAM1 receptors.
PMID 25824372
Abstract
Carcinoembryonicantigen-related cell adhesion molecule 1 (CEACAM1) is a receptor involved in the regulation of NK-cell function. In most species, the CEACAM1 cytoplasmic tail possesses a membrane-proximal ITIM paired with a membrane-distal immunoreceptor tyrosine-based switch motif (ITSM) signaling motif. Human CEACAM1 has phylogenetically relatively recently acquired a second ITIM instead of the ITSM and was shown to inhibit NKG2D-mediated NK-cell activation. Here, we compare the function of bovine and human CEACAM1. We show that in addition to NKG2D, human CEACAM1 can inhibit NK-cell activation via NKp30 or 2B4. Bovine CEACAM1, possessing an ITIM and an ITSM signaling motif, is also inhibitory. However, bovine CEACAM1 inhibition of NKp30-mediated lysis is less pronounced compared with its human counterpart. Bovine CEACAM1 inhibition is dependent on the membrane-proximal ITIM and our data suggest that also the membrane distal ITSM motif contributes to inhibitory signaling. Biochemically, human and bovine CEACAM1 can recruit the phosphatases SHP-1 and SHP-2 after receptor phosphorylation to a similar extend. Bovine CEACAM1 can additionally recruit the adapter molecule Ewing's sarcoma virus-activated transcript-2 (EAT-2), but not SLAM-associated protein (SAP). Taken together, we show that although human and bovine CEACAM1 are differentially equipped with ITIM and ITSM motifs, both receptors can inhibit NKp30 and 2B4 activation of NK cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV41962
Anti-APCS antibody produced in rabbit, IgG fraction of antiserum
HPA053294
Anti-APCS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501094
Anti-NCR3 antibody produced in rabbit, affinity isolated antibody
SAB1412940
Anti-NCR3 antibody produced in rabbit, purified immunoglobulin, 1 mg/mL
SAB2101628
Anti-NR0B2 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101629
Anti-NR0B2 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2101630
Anti-NR0B2 (ab3) antibody produced in rabbit, affinity isolated antibody
SAB2101631
Anti-NR0B2 (ab4) antibody produced in rabbit, affinity isolated antibody
SAB1406664
Anti-NR0B2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2700022
Anti-NS1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700186
Anti-NS1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700193
Anti-NS1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300559
Anti-PTPN11 (Ab-542) antibody produced in rabbit, affinity isolated antibody
SAB4300560
Anti-PTPN11 (Ab-580) antibody produced in rabbit, affinity isolated antibody
SAB2900281
Anti-PTPN11 (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900375
Anti-PTPN11 (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
GW21495A Anti-PTPN11 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1406345
Anti-PTPN11 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4300558
Anti-PTPN6 (Ab-536) antibody produced in rabbit, affinity isolated antibody
SAB1406344
Anti-PTPN6 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001466
Anti-PTPN6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW21487 Anti-PTPNS1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1306039
ANTI-SAP (CENTER E300) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4502536
Anti-SH-PTP2 antibody produced in rabbit, affinity isolated antibody
SAB1406096
Anti-SH2D1A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410764
Anti-SH2D1A antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1408257
Anti-SH2D1B antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401969
Anti-SH2D1B antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA054791 Anti-SH2D1B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502529
Anti-SHP-1, C-Terminal antibody produced in rabbit, affinity isolated antibody