EMAIL THIS PAGE TO A FRIEND
The American journal of tropical medicine and hygiene
Dengue serotype-specific differences in clinical manifestation, laboratory parameters and risk of severe disease in adults, singapore.
PMID 25825386
Abstract
Studies on serotype-specific features of dengue and disease severity on adults are limited. We prospectively recruited adult febrile patients without alternate diagnosis to dengue from April 2005 to December 2011. Outcomes were defined using both the World Health Organization (WHO) 1997 and 2009 criteria; Dengue hemorrhagic fever (DHF) and severe dengue (SD). Infecting serotype was identified in 469 dengue-confirmed patients comprising 22.0% dengue virus serotype 1 (DENV-1), 57.1% DENV-2, 17.1% DENV-3, and 3.8% DENV-4. Cases infected with DENV-1 were more likely to present with red eyes whereas presence of joint pain and lower platelet count was associated with DENV-2 cases. After adjusting for potential confounders, DENV-1 was associated with both DHF (adjusted Relative Risk [aRR] = 1.74) and SD (aRR = 2.1) whereas DENV-2 had a lower risk of DHF (aRR = 0.5). DENV-1 genotype 1 and DENV-2 cosmopolitan were the predominant genotypes identified. Infecting dengue serotype and possibly genotype may play an important role in disease severity among adult dengue patients in Singapore.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RAB0806 Mouse IgM ELISA Kit