EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Sca-1+Lin-CD117- mesenchymal stem/stromal cells induce the generation of novel IRF8-controlled regulatory dendritic cells through Notch-RBP-J signaling.
PMID 25825436
Abstract
Mesenchymal stem/stromal cells (MSCs) can influence the destiny of hematopoietic stem/progenitor cells (HSCs) and exert broadly immunomodulatory effects on immune cells. However, how MSCs regulate the differentiation of regulatory dendritic cells (regDCs) from HSCs remains incompletely understood. In this study, we show that mouse bone marrow-derived Sca-1(+)Lin(-)CD117(-) MSCs can drive HSCs to differentiate into a novel IFN regulatory factor (IRF)8-controlled regDC population (Sca(+) BM-MSC-driven DC [sBM-DCs]) when cocultured without exogenous cytokines. The Notch pathway plays a critical role in the generation of the sBM-DCs by controlling IRF8 expression in an RBP-J-dependent way. We observed a high level of H3K27me3 methylation and a low level of H3K4me3 methylation at the Irf8 promoter during sBM-DC induction. Importantly, infusion of sBM-DCs could alleviate colitis in mice with inflammatory bowel disease by inhibiting lymphocyte proliferation and increasing the numbers of CD4(+)CD25(+) regulatory T cells. Thus, these data infer a possible mechanism for the development of regDCs and further support the role of MSCs in treating immune disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV36912
Anti-ASCL2 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV36913
Anti-ASCL2 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1305026
ANTI-ASCL2 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2104130
Anti-ASCL2 antibody produced in rabbit, affinity isolated antibody
SAB2106440
Anti-ASCL2 antibody produced in rabbit, affinity isolated antibody
SAB2100164
Anti-ASH2L (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100165
Anti-ASH2L (ab2) antibody produced in rabbit, affinity isolated antibody
AV34203
Anti-ASH2L antibody produced in rabbit, affinity isolated antibody
SAB4300685
Anti-ASH2L antibody produced in rabbit, affinity isolated antibody
HPA042289
Anti-ASH2L antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2900054
Anti-EPHA4 (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900055
Anti-EPHA4 (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
E6900 Anti-EphA4 antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB2107644
Anti-EPHA4 antibody produced in rabbit, affinity isolated antibody
HPA002267
Anti-IRF8 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV100841
Anti-IRF8 antibody produced in rabbit, affinity isolated antibody
HPA002531
Anti-IRF8 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103913
Anti-IRF8 antibody produced in rabbit, affinity isolated antibody
SAB2107842
Anti-IRF8 antibody produced in rabbit, affinity isolated antibody
SAB1300326 Anti-MLL1 (C-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4300387
Anti-RB1 (Ab-780) antibody produced in rabbit, affinity isolated antibody
SAB4300385
Anti-RB1 (Ab-795) antibody produced in rabbit, affinity isolated antibody
SAB4300386
Anti-RB1 (Ab-807) antibody produced in rabbit, affinity isolated antibody
AV33212
Anti-RB1 antibody produced in rabbit, affinity isolated antibody
S5313
Anti-Sir2 (AS-16) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
S5322
Anti-Sirt1 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB3500493
Anti-SIRT1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501994
Anti-SIRT1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
S5447
Anti-SIRT1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA006295
Anti-SIRT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution