EMAIL THIS PAGE TO A FRIEND
Oncotarget
HPV16 E5 deregulates the autophagic process in human keratinocytes.
PMID 25826082
Abstract
Autophagy plays key roles during host defense against pathogens, but viruses have evolved strategies to block the process or to exploit it for replication and successful infection. The E5 oncoprotein of human papillomavirus type 16 (HPV16 E5) perturbs epithelial homeostasis down-regulating the expression of the keratinocyte growth factor receptor (KGFR/FGFR2b), whose signaling induces autophagy. Here we investigated the possible effects of 16E5 on autophagy in human keratinocytes expressing the viral protein. The 16E5 presence strongly inhibited the autophagic process, while forced expression and activation of KGFR counteracted this effect, demonstrating that the viral protein and the receptor exert opposite and interplaying roles not only on epithelial differentiation, but also in the control of autophagy. In W12 cells, silencing of the 16E5 gene in the context of the viral full length genome confirmed its role on autophagy inhibition. Finally, molecular approaches showed that the viral protein interferes with the transcriptional regulation of autophagy also through the impairment of p53 function, indicating that 16E5 uses parallel mechanisms for autophagy impairment. Overall our results further support the hypothesis that a transcriptional crosstalk among 16E5 and KGFR might be the crucial molecular driver of epithelial deregulation during early steps of HPV infection and transformation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2500395
Anti-FGFR2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1412031
ANTI-FGFR2 antibody produced in mouse, clone 3F4, purified immunoglobulin, buffered aqueous solution
HPA035305
Anti-FGFR2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500889
Anti-FGFR2 antibody produced in rabbit, affinity isolated antibody
SAB3701033 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Texas Red™ antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701019 Anti-Mouse IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed-Texas Red™ antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700996 Anti-Mouse IgG F(ab′)2-Texas Red™ antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB1406748
Anti-SQSTM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB3500430
Anti-SQSTM1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2104334
Anti-SQSTM1 antibody produced in rabbit, affinity isolated antibody
SAB3500594
Anti-ZIP3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
EHU125631 MISSION® esiRNA, esiRNA human FGFR2 (esiRNA1)
EHU101411 MISSION® esiRNA, esiRNA human ARHGEF15 (esiRNA1)
EMU079351 MISSION® esiRNA, esiRNA targeting mouse Arhgef15 (esiRNA1)
WH0002263M1
Monoclonal Anti-FGFR2 antibody produced in mouse, clone 1G3, purified immunoglobulin, buffered aqueous solution
SAB1403815
Monoclonal Anti-FGFR2 antibody produced in mouse, clone 2C11, purified immunoglobulin, buffered aqueous solution
WH0008878M1
Monoclonal Anti-SQSTM1 antibody produced in mouse, clone 2C11, purified immunoglobulin, buffered aqueous solution
SAB1404647
Monoclonal Anti-SQSTM1 antibody produced in mouse, clone 1C9, purified immunoglobulin, buffered aqueous solution