EMAIL THIS PAGE TO A FRIEND
The Journal of investigative dermatology
Retinoids Bias Integrin Expression and Function in Cutaneous T-Cell Lymphoma.
PMID 25826424
Abstract
Cutaneous T-cell lymphoma (CTCL) is a heterogeneous group of malignancies characterized by accumulation of malignant T-cells within the skin. Retinoids, metabolic derivatives, and synthetic analogs of vitamin A embody an effective CTCL therapy with over three decades of clinical use. The established mechanism of action is induction of growth arrest and apoptosis. However, the natural role of retinoids in T-cell biology is imprinting gut-homing properties by inducing integrin α4β7 expression. How the natural role of retinoids relates to therapeutic effectiveness in CTCL has not been addressed and merits investigation. Here we provide evidence that retinoids, including Bexarotene, selectively induce CTCL lineages to increase integrin β7 expression and function prior to growth arrest and apoptosis. Interestingly, augmented CTCL cell adhesion obtained with retinoid exposure was potently attenuated by 1,25-dihydroxyvitamin D3, a metabolic vitamin derivative involved in prompting immune cell skin homing. The integrin-dependent adhesion changes in CTCL cells occurred through synergistic activation of RAR and RXR nuclear receptors. These data explore the early cellular changes induced by retinoids that may be pivotal to sensitizing CTCL cells to growth arrest and apoptosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0282
Bexarotene, ≥98% (HPLC)
C24H28O2
S8951
SR11237, ≥98% (HPLC)
C24H28O4
T3757
TTNPB
C24H28O2
V9378
Vitamin K2
C31H40O2