EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Nanoscale metallogel via self-assembly of self-assembled trinuclear coordination rings: multi-stimuli-responsive soft materials.
PMID 25826610
Abstract
A rare variety of coordination rings having the M3L6 composition are prepared by the combination of Pd(NO3)2 with an imidazolyl or benzimidazolyl appended bidentate non-chelating ligand (i.e.L1 or L2). The variable concentration 1H NMR spectra of the trinuclear complexes [Pd3(L1)6](NO3)6, 1a and [Pd3(L2)6](NO3)6, 2a in DMSO-d6 provided valuable information on the self-assembly phenomenon of the already self-assembled complexes. Interestingly, the signals of 2a are broadened upon increasing the concentration. The solution of 2a in DMSO formed a supramolecular metallogel above a certain concentration (i.e. 2% w/v), however, complex 1a could not form any gel. This phenomenon is attributed to the presence of an auxiliary π-surface in the benzimidazole moiety in L2, in contrast to the imidazole moiety of L1. The influence of anions in gel formation was studied by preparing several samples using a variety of palladium(II) salts and L2 in DMSO. It was found that oxoanions like nitrate, perchlorate, triflate and tosylate are amicable for gel formation, probably due to their capabilities of forming H-bonds. The counter anions like tetrafluoroborate, hexafluorophosphate, or hexafluoroantimonate could not assist in the formation of gel. The stimuli-responsive nature of the gel and the reversible gel–sol conversion were demonstrated by dis-assembly and re-assembly processes of 2a as controlled by a pair of stimuli such as halide-nitrate, DMAP-HNO3, and ethylenediamine-Pd(NO3)2. No gel could be prepared by the combination of Ni(NO3)2 or Pt(NO3)2 with ligand L2. Thus, a subtle change in the ligand design, metal ion and counter anion is demonstrated as the responsible parameter for the construction of the three component multi-stimuli-responsive supramolecular gel.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
107700
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
C7H10N2
714844
4-(Dimethylamino)pyridine solution, 0.5 M in THF
C7H10N2
714720
4-(Dimethylamino)pyridine solution, 0.5 M in ethyl acetate
C7H10N2
700169
4-(Dimethylamino)pyridine, ChemDose tablets, Loading: 0.04 mmol tablet.
C7H10N2
194123
Benzimidazole, 98%
C7H6N2
03140
Copper(I) iodide, purum, ≥99.5%
CuI
215554
Copper(I) iodide, 99.999% trace metals basis
CuI
792063
Copper(I) iodide, anhydrous, 99.995% trace metals basis
CuI
792527
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
C3H4N2
I5513
Imidazole, for molecular biology, ≥99% (titration)
C3H4N2
I0250
Imidazole, ≥99% (titration), crystalline
C3H4N2
I2399
Imidazole, ACS reagent, ≥99% (titration)
C3H4N2
56750
Imidazole, puriss. p.a., ≥99.5% (GC)
C3H4N2
I202
Imidazole, ReagentPlus®, 99%
C3H4N2
56748
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
C3H4N2
56749
Imidazole, BioUltra, ≥99.5% (GC)
C3H4N2
68268
Imidazole buffer Solution, BioUltra, 1 M in H2O
C3H4N2
D1156
N,N-Dimethylglycine, ≥99%
C4H9NO2
S8157
Silver nitrate, BioXtra, >99% (titration)
AgNO3
S7276
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
AgNO3
S1179
Silver nitrate, meets USP testing specifications
AgNO3
204390
Silver nitrate, 99.9999% trace metals basis
AgNO3
792276
Silver nitrate, anhydrous, 99.999% trace metals basis
AgNO3
248762
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
AgNO3
85193
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
AgNO3