EMAIL THIS PAGE TO A FRIEND
Nature communications
A muscle-liver-fat signalling axis is essential for central control of adaptive adipose remodelling.
PMID 25827749
Abstract
Skeletal muscle has a pleiotropic role in organismal energy metabolism, for example, by storing protein as an energy source, or by excreting endocrine hormones. Muscle proteolysis is tightly controlled by the hypothalamus-pituitary-adrenal signalling axis via a glucocorticoid-driven transcriptional programme. Here we unravel the physiological significance of this catabolic process using skeletal muscle-specific glucocorticoid receptor (GR) knockout (GRmKO) mice. These mice have increased muscle mass but smaller adipose tissues. Metabolically, GRmKO mice show a drastic shift of energy utilization and storage in muscle, liver and adipose tissues. We demonstrate that the resulting depletion of plasma alanine serves as a cue to increase plasma levels of fibroblast growth factor 21 (FGF21) and activates liver-fat communication, leading to the activation of lipolytic genes in adipose tissues. We propose that this skeletal muscle-liver-fat signalling axis may serve as a target for the development of therapies against various metabolic diseases, including obesity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
166898
3-Hydroxybutyric acid, 95%
C4H8O3
C9033
β-Chloro-L-alanine hydrochloride, Alanine aminotransferase inhibitor
C3H6ClNO2 · HCl
T1378
Sigma 7-9®, ≥99% (titration), crystalline
C4H11NO3
154563
Tris(hydroxymethyl)aminomethane, ultrapure grade, ≥99.9%
C4H11NO3
252859
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
C4H11NO3
RDD008
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
C4H11NO3
T6066
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, cell culture tested, ≥99.9% (titration)
C4H11NO3
T6791
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
C4H11NO3
T4661
Trizma® base, ≥99.9% (titration), crystalline
C4H11NO3
93362
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
C4H11NO3
93352
Trizma® base, ≥99.0% (T)
C4H11NO3
93350
Trizma® base, puriss. p.a., ≥99.7% (T)
C4H11NO3
33742
Trizma® base, puriss. p.a., buffer substance, ≥99.5%
C4H11NO3
T6687
Tromethamine, meets USP testing specifications
C4H11NO3
RES3137T-A7
Tromethamine, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., suitable for cell culture, meets USP testing specifications
C4H11NO3
RES3193T-A7
Tromethamine, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture, Meets USP, EP, JPC, BP testing specifications.
C4H11NO3