EMAIL THIS PAGE TO A FRIEND
Anaesthesia
Effect of magnesium sulphate on sugammadex reversal time for neuromuscular blockade: a randomised controlled study.
PMID 25829048
Abstract
Magnesium potentiates neuromuscular blockade. Sugammadex reverses rocuronium-induced blockade. The aim of this study was to determine the effect of pre-treatment with magnesium sulphate on sugammadex reversal time for neuromuscular blockade. Seventy-three patients were randomly assigned to receive magnesium sulphate (40 mg.kg(-1) ) or saline intravenously. After anaesthetic induction, continuous train-of-four monitoring was performed and rocuronium was administered (0.6 mg.kg(-1) ). When a second twitch appeared, the patients received sugammadex (2 mg.kg(-1) ). The median (IQR [range]) reversal time of moderate neuromuscular blockade to a train-of-four ratio of 0.9 facilitated by sugammadex was 115 (93-177.5 [68-315]) s in the magnesium group and 120 (105-140 [70-298]) s in the saline group (p = 0.79). The median (IQR [range]) clinical duration was 45 (35.5-53 [22-102]) min in the magnesium group and 37 (31-43 [19-73]) min in the saline group (p = 0.031). Pre-treatment with magnesium did not significantly affect sugammadex reversal time of moderate neuromuscular blockade induced by rocuronium.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D126608
2,6-Diisopropylphenol, 97%
C12H18O