EMAIL THIS PAGE TO A FRIEND
PLoS pathogens
A strategy for O-glycoproteomics of enveloped viruses--the O-glycoproteome of herpes simplex virus type 1.
PMID 25830354
Abstract
Glycosylation of viral envelope proteins is important for infectivity and interaction with host immunity, however, our current knowledge of the functions of glycosylation is largely limited to N-glycosylation because it is difficult to predict and identify site-specific O-glycosylation. Here, we present a novel proteome-wide discovery strategy for O-glycosylation sites on viral envelope proteins using herpes simplex virus type 1 (HSV-1) as a model. We identified 74 O-linked glycosylation sites on 8 out of the 12 HSV-1 envelope proteins. Two of the identified glycosites found in glycoprotein B were previously implicated in virus attachment to immune cells. We show that HSV-1 infection distorts the secretory pathway and that infected cells accumulate glycoproteins with truncated O-glycans, nonetheless retaining the ability to elongate most of the surface glycans. With the use of precise gene editing, we further demonstrate that elongated O-glycans are essential for HSV-1 in human HaCaT keratinocytes, where HSV-1 produced markedly lower viral titers in HaCaT with abrogated O-glycans compared to the isogenic counterpart with normal O-glycans. The roles of O-linked glycosylation for viral entry, formation, secretion, and immune recognition are poorly understood, and the O-glycoproteomics strategy presented here now opens for unbiased discovery on all enveloped viruses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA011008
Anti-GOLGB1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100948
Anti-GOLGB1 antibody produced in rabbit, affinity isolated antibody
HPA011555
Anti-GOLGB1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
G4420 Anti-GRP94 (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4503031
Anti-GRP94, C-Terminal antibody produced in rabbit, affinity isolated antibody
HPA003901
Anti-HSP90B1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA008424
Anti-HSP90B1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV40464
Anti-HSP90B1 antibody produced in rabbit, IgG fraction of antiserum
SAB2101094
Anti-HSP90B1 antibody produced in rabbit, affinity isolated antibody
A1293 Anti-Mouse IgG (Fab specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A2179 Anti-Mouse IgG (Fab specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A1682 Anti-Mouse IgG (Fab specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B7151 Anti-Mouse IgG (Fab specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B0529 Anti-Mouse IgG (Fab specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
T6528
Anti-Mouse IgG (Fab specific)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
T7782 Anti-Mouse IgG (Fab specific)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701030 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701031 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701028 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Fluorescein antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701029 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701011 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701033 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Texas Red™ antibody produced in goat, affinity isolated antibody, lyophilized powder
A1418 Anti-Mouse IgG (Fc specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A2429 Anti-Mouse IgG (Fc specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A7434
Anti-Mouse IgG (Fc specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
B7401 Anti-Mouse IgG (Fc specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B9904 Anti-Mouse IgG (Fc specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A3438 Anti-Mouse IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
A1047 Anti-Mouse IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B7022 Anti-Mouse IgG (γ-chain specific)−Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution