EMAIL THIS PAGE TO A FRIEND
International journal of phytoremediation
Effects of Amendments on Growth and Uptake of Cd and Zn by Wetland Plants, Typha angustifolia and Colocasia esculenta from Contaminated Sediments.
PMID 25831275
Abstract
A pot study was conducted to compare the effects of amendments (CaHPO4 and cow manure) on growth and uptake of Cd and Zn from contaminated sediments by two wetland plant species, Typha angustifolia and Colocasia esculenta. Contaminated sediments (Cd 33.2 mg kg(-1) and Zn 363 mg kg(-1)) were collected from Mae Tao basin, Mae Sot district, Tak province, Thailand. The experiment consisted of 4 treatments: control (uncontaminated sediment), Cd/Zn, Cd/Zn + 5% CaHPO4, and Cd/Zn + 10% cow manure. Plants were grown for 3 months in the greenhouse. The addition of CaHPO4 resulted in the highest relative growth rate (RGR) and highest Cd accumulation in both T. angustifolia and C. esculenta while the lowest RGR was found in C. esculenta grown in the cow manure treatment. Both plant species had higher concentrations of metals (Cd, Zn) in their belowground parts. None of the amendments affected Zn accumulation. C. esculenta exhibited the highest uptake of both Cd and Zn. The results clearly demonstrated the phytoremediation potential of C. esculenta and the enhancement of this potential by CaHPO4 amendment.