EMAIL THIS PAGE TO A FRIEND
The Journal of veterinary medical science
Stromal cell-derived factor-1 (SDF1)-dependent recruitment of bone marrow-derived renal endothelium-like cells in a mouse model of acute kidney injury.
PMID 25833353
Abstract
Ischemic acute kidney injury (AKI) is the most key pathological event for accelerating progression to chronic kidney disease through vascular endothelial injury or dysfunction. Thus, it is critical to elucidate the molecular mechanism of endothelial protection and regeneration. Emerging evidence indicates that bone marrow-derived cells (BMCs) contribute to tissue reconstitution in several types of organs post-injury, but little is known whether and how BMCs contribute to renal endothelial reconstitution, especially in an early-stage of AKI. Using a mouse model of ischemic AKI, we provide evidence that incorporation of BMCs in vascular components (such as endothelial and smooth muscle cells) becomes evident within four days after renal ischemia and reperfusion, associated with an increase in stromal cell-derived factor-1 (SDF1) in endothelium and that in CXCR4/SDF1-receptor in BMCs. Notably, anti-CXCR4 antibody decreased the numbers of infiltrated BMCs and BMC-derived endothelium-like cells, but not of BMC-derived smooth muscle cell-like cells. These results suggest that reconstitution of renal endothelium post-ischemia partially depends on a paracrine loop of SDF1-CXCR4 between resident endothelium and BMCs. Such a chemokine ligand-receptor system may be attributable for selecting a cellular lineage (s), required for renal vascular protection, repair and homeostasis, even in an earlier phase of AKI.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1165
Pimonidazole, ≥98% (HPLC)
C11H18N4O3