EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Leucine-rich repeat kinase 2 modulates neuroinflammation and neurotoxicity in models of human immunodeficiency virus 1-associated neurocognitive disorders.
PMID 25834052
Abstract
Leucine-rich repeat kinase 2 (LRRK2) is the single most common genetic cause of both familial and sporadic Parkinson's disease (PD), both of which share pathogenetic and neurologic similarities with human immunodeficiency virus 1 (HIV-1)-associated neurocognitive disorders (HAND). Pathologic LRRK2 activity may also contribute to neuroinflammation, because microglia lacking LRRK2 exposed to proinflammatory stimuli have attenuated responses. Because microglial activation is a hallmark of HIV-1 neuropathology, we have investigated the role of LRRK2 activation using in vitro and in vivo models of HAND. We hypothesize that LRRK2 is a key modulator of microglial inflammatory responses, which play a pathogenic role in both HAND and PD, and that these responses may cause or exacerbate neuronal damage in these diseases. The HIV-1 Tat protein is a potent neurotoxin produced during HAND that induces activation of primary microglia in culture and long-lasting neuroinflammation and neurotoxicity when injected into the CNS of mice. We found that LRRK2 inhibition attenuates Tat-induced pS935-LRRK2 expression, proinflammatory cytokine and chemokine expression, and phosphorylated p38 and Jun N-terminal kinase signaling in primary microglia. In our murine model, cortical Tat injection in LRRK2 knock-out (KO) mice results in significantly diminished neuronal damage, as assessed by microtubule-associated protein 2 (MAP2), class III β-tubulin TUJ1, synapsin-1, VGluT, and cleaved caspase-3 immunostaining. Furthermore, Tat-injected LRRK2 KO animals have decreased infiltration of peripheral neutrophils, and the morphology of microglia from these mice were similar to that of vehicle-injected controls. We conclude that pathologic activation of LRRK2 regulates a significant component of the neuroinflammation associated with HAND.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB377
Anti-NeuN Antibody, clone A60, 500 µg, Anti-NeuN Antibody, clone A60 detects level of NeuN and has been published and validated for use in FC, IC, IF, IH, IH(P), IP and WB.
AB5905
Anti-Vesicular Glutamate Transporter 1 Antibody, 50 µL, This Anti-Vesicular Glutamate Transporter 1 Antibody is validated for use in Immunohistochemistry for the detection of Vesicular Glutamate Transporter 1 with more than 55 product citations.
A3294
Bovine Serum Albumin, heat shock fraction, protease free, pH 7, ≥98%
94563
Dimethyl sulfoxide, analytical standard
C2H6OS
01934
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
C2H6OS
1211006
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
C2H6OS
D2050000
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
C2H6OS
1295607
Glycerin, United States Pharmacopeia (USP) Reference Standard
C3H8O3
PHR1020
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O3
49779
Glycerol, tested according to Ph.Eur., anhydrous
C3H8O3
77067
Glycerol, analytical standard
C3H8O3
12146
Glycerol 3 M solution, 3 M
C3H8O3 · xH2O
76524
Glycine, certified reference material, TraceCERT®
C2H5NO2
50049
Glycine, BioUltra, ≥99.0% (NT)
C2H5NO2
50050
Glycine, ≥99.0% (NT)
C2H5NO2
50058
Glycine, tested according to Ph.Eur.
C2H5NO2
PHR1111
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
C2H5NO2
1295800
Glycine, United States Pharmacopeia (USP) Reference Standard
C2H5NO2
94119
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
C2H5NO2
G0450000
Glycine, European Pharmacopoeia (EP) Reference Standard
C2H5NO2
P1149
Poly-D-lysine hydrobromide, mol wt 150,000-300,000
71383
Sodium chloride, tested according to Ph.Eur.
ClNa
71387
Sodium chloride, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
ClNa
PHR1321
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
ClNa
71177
Sodium standard for AAS, ready-to-use, traceable to BAM, in H2O
ClNa