EMAIL THIS PAGE TO A FRIEND
British journal of anaesthesia
Intrajugular balloon catheter reduces air embolism in vitro and in vivo.
PMID 25835025
Abstract
Neurosurgical procedures requiring a sitting position may put the patient at risk of a potentially life-threatening air embolism. Transient manual jugular venous compression limits further air entry in this situation. This study presents an alternative technique aimed at reducing the risk of air embolism. In an in vitro model, an intrajugular balloon catheter was inserted to demonstrate that this device prevents air embolism. In an in vivo study, this device was bilaterally placed into jugular vessels in pigs. Using an ultrasound technique, blood flow was monitored and jugular venous pressure was recorded before and during cuff inflation. Air was applied proximally to the inflated cuffs to test the hypothesis that this novel device blocks air passage. In vitro, the intrajugular balloon catheter reliably prevented further air entry (n=10). Additionally, accumulated air could be aspirated from an orifice of the catheter (n=10). In vivo, inflation of the catheter balloon completely obstructed venous blood flow (n=8). Bilateral inflation of the cuff significantly increased the proximal jugular venous pressure from 9.8 (2.4) mm Hg to 14.5 (2.5) mm Hg (n=8, P<0.05). Under conditions mimicking an air embolism, air passage across the inflated cuffs was prevented and 78 (20%) (n=6) of the air dose could be aspirated by the proximal orifice of the catheter. These findings may serve as a starting point for the development of intrajugular balloon catheters designed to reduce the risk of air embolism in patients undergoing neurosurgery in a sitting position.