EMAIL THIS PAGE TO A FRIEND
Oncotarget
A novel role for drebrin in regulating progranulin bioactivity in bladder cancer.
PMID 25839164
Abstract
We recently established a critical role for the growth factor progranulin in bladder cancer insofar as progranulin promotes urothelial cancer cell motility and contributes, as an autocrine growth factor, to the transformed phenotype by modulating invasion and anchorage-independent growth. In addition, progranulin expression is upregulated in invasive bladder cancer tissues compared to normal controls. However, the molecular mechanisms of progranulin action in bladder cancer have not been fully elucidated. In this study, we searched for novel progranulin-interacting proteins using pull-down assays with recombinant progranulin and proteomics. We discovered that drebrin, an F-actin binding protein, bound progranulin in urothelial cancer cells. We characterized drebrin function in urothelial cancer cell lines and showed that drebrin is critical for progranulin-dependent activation of the Akt and MAPK pathways and modulates motility, invasion and anchorage-independent growth. In addition, drebrin regulates tumor formation in vivo and its expression is upregulated in bladder cancer tissues compared to normal tissue controls. Our data are translationally relevant as indicate that drebrin exerts an essential functional role in the regulation of progranulin action and may constitute a novel target for therapeutic intervention in bladder tumors. In addition, drebrin may serve as novel biomarker for bladder cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2700952
Anti-DBN1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701104
Anti-DBN1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
D3816
Anti-Drebrin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4200310
Anti-Granulin (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB4200557
Anti-Granulin antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
SAB2501196
Anti-GRN antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502030
Anti-GRN antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA008763
Anti-GRN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA028747 Anti-GRN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410570
Anti-GRN antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1402168
Monoclonal Anti-DBN1 antibody produced in mouse, clone 2E11, purified immunoglobulin, buffered aqueous solution
WH0002896M1
Monoclonal Anti-GRN antibody produced in mouse, clone 1F5, purified immunoglobulin, buffered aqueous solution
SAB1403884
Monoclonal Anti-GRN, (C-terminal) antibody produced in mouse, clone 1A8, purified immunoglobulin, buffered aqueous solution