Description
Molecular Formula
Add to Cart
SAB1405458
Anti-ANXA3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1305642
ANTI-APG5 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
A0731
Anti-ATG5 (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody
A0856
Anti-ATG5 (N-terminal) antibody produced in rabbit, affinity isolated antibody, PBS solution
GW22210F
Anti-ATG5 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500448
Anti-ATG5 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA042973
Anti-ATG5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1407006
Anti-ATG7 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
A2856
Anti-ATG7 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA007639
Anti-ATG7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410101
Anti-ATG7 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2701263
Anti-DCTN4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2107220
Anti-DCTN4 antibody produced in rabbit, affinity isolated antibody
SAB2105017
Anti-DCTN4, (N-terminal) antibody produced in rabbit, affinity isolated antibody
GW22005A Anti-GTF2H1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22005B Anti-GTF2H1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV100811
Anti-GTF2H1 antibody produced in rabbit, IgG fraction of antiserum
SAB2104463
Anti-GTF2H1 antibody produced in rabbit, affinity isolated antibody
SAB1306269
ANTI-LC3 (APG8A/B) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW22431A Anti-NUP62 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA005435
Anti-NUP62 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101664
Anti-NUP62 antibody produced in rabbit, affinity isolated antibody
SAB1410438
Anti-NUP62 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
WH0000306M12
Monoclonal Anti-ANXA3 antibody produced in mouse, clone 4F1, purified immunoglobulin, buffered aqueous solution
SAB1403553
Monoclonal Anti-ANXA3 antibody produced in mouse, clone 1E2, purified immunoglobulin, buffered aqueous solution
SAB4100143
Monoclonal Anti-ANXA3 antibody produced in mouse, clone ANXA3-A107, culture supernatant
WH0009474M1
Monoclonal Anti-ATG5 antibody produced in mouse, clone 3D2, purified immunoglobulin, buffered aqueous solution
A2859
Monoclonal Anti-ATG5 antibody produced in mouse, clone ATG5-18, purified immunoglobulin, buffered aqueous solution
SAB1404726
Monoclonal Anti-ATG5, (C-terminal) antibody produced in mouse, clone 1C4, purified immunoglobulin, buffered aqueous solution
SAB4200304
Monoclonal Anti-Atg7 antibody produced in mouse, ~1.0 mg/mL, clone ATG7-13, purified immunoglobulin