Description
Molecular Formula
Add to Cart
SAB4501330
Anti-GSK3 antibody produced in rabbit, affinity isolated antibody
SAB4300287
Anti-GSK3B (Ab-9) antibody produced in rabbit, affinity isolated antibody
SAB2100978
Anti-GSK3B antibody produced in rabbit, affinity isolated antibody
HPA028017
Anti-GSK3B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA017736
Anti-HS3ST2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502298
Anti-PP2A-α, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1300599
Anti-PPP2R1A (C-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1300598
Anti-PPP2R1A (N-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1401296
Anti-PPP2R1A antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2103131
Anti-PPP2R1A, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2501263
Anti-PPP2R4 (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406288
Anti-PPP2R4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA005695
Anti-PPP2R4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401298
Anti-PPP2R4 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
CKOZFN10307 CompoZr® Knockout ZFN Kit, Human HS3ST2 (NM_006043)
WH0002932M4
Monoclonal Anti-GSK3B antibody produced in mouse, clone 1H3, purified immunoglobulin, buffered aqueous solution
SAB5300175 Monoclonal Anti-GSK3B antibody produced in mouse, clone 3D10, ascites fluid
SAB1401297
Monoclonal Anti-PPP2R1A antibody produced in mouse, clone 4E6, purified immunoglobulin, buffered aqueous solution
SAB5300403
Monoclonal Anti-PPP2R4 antibody produced in mouse, clone 4D9, purified immunoglobulin, buffered aqueous solution