EMAIL THIS PAGE TO A FRIEND
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics
Interaction Studies of Resolvin E1 Analog (RX-10045) with Efflux Transporters.
PMID 25844889
Abstract
Screening interactions of a resolvin E1 analog (RX-10045) with efflux transporters (P-glycoprotein [P-gp], multidrug resistance-associated protein [MRP2], and breast cancer-resistant protein [BCRP]). Madin-Darby canine kidney cells transfected with P-gp, MRP2, and BCRP genes were selected for this study. [3H]-Digoxin, [3H]-vinblastine and [3H]-abacavir were selected as model substrates for P-gp, MRP2, and BCRP. Uptake and permeability studies across cell monolayer in both apical to basal (AP-BL) and BL-AP of these substrates were conducted in the presence of specific efflux pump inhibitors and RX-10045. Cell viability studies were conducted with increasing concentrations of RX-10045. Uptake studies showed a higher accumulation in the presence of inhibitors (GF120918 and ketoconazole for P-gp; MK571 for MRP2; and β-estradiol for BCRP) as well as RX-10045. Similarly, dose-dependent inhibition studies demonstrated higher accumulation of various substrates ([3H]-digoxin, [3H]-vinblastine, and [3H]-abacavir) in the presence of RX-10045. IC50 values of dose-dependent inhibition of RX-10045 for P-gp, MRP2, and BCRP were 239±11.2, 291±79.2, and 300±42 μM, respectively. Cell viability assay indicated no apparent toxicity up to 350 μM concentration. Enhanced permeability for model substrates was observed in the presence of RX-10045. Uptake studies in human corneal epithelial cells suggest that RX-10045 is a strong inhibitor of organic cation transporter-1 (OCT-1). In summary, the resolvin analog (RX-10045) was identified as a substrate/inhibitor for efflux transporters (MRP2 and BCRP). Also, RX-10045 appears to be a strong inhibitor/substrate of OCT-1. Novel formulation strategies such as nanoparticles, nanomicelles, and liposomes for circumventing efflux barriers and delivering higher drug concentrations leading to a higher therapeutic efficacy may be employed.