EMAIL THIS PAGE TO A FRIEND
Clinical and experimental immunology
Remission of systemic lupus erythematosus disease activity with regulatory cytokine interleukin (IL)-35 in Murphy Roths Large (MRL)/lpr mice.
PMID 25845911
Abstract
The immunological mechanisms mediated by regulatory cytokine interleukin (IL)-35 are unclear in systemic lupus erythematosus (SLE). We investigated the frequency of CD4(+) CD25(+) forkhead box protein 3 (FoxP3)(+) regulatory T (Treg ) and IL-10(+) regulatory B (Breg ) cells and related immunoregulatory mechanisms in a female Murphy Roths Large (MRL)/lpr mouse model of spontaneous lupus-like disease, with or without IL-35 treatment. A remission of histopathology characteristics of lupus flare and nephritis was observed in the MRL/lpr mice upon IL-35 treatment. Accordingly, IL-35 and IL-35 receptor subunits (gp130 and IL-12Rβ2) and cytokines of MRL/lpr and BALB/c mice (normal controls) were measured. The increased anti-inflammatory cytokines and decreased proinflammatory cytokines were possibly associated with the restoration of Treg and Breg frequency in MRL/lpr mice with IL-35 treatment, compared to phosphate-buffered saline (PBS) treatment. mRNA expressions of Treg -related FoxP3, IL-35 subunit (p35 and EBI3) and soluble IL-35 receptor subunit (gp130 and IL12Rβ2) in splenic cells were up-regulated significantly in IL-35-treated mice. Compared with the PBS treatment group, IL-35-treated MRL/lpr mice showed an up-regulation of Treg -related genes and the activation of IL-35-related intracellular Janus kinase/signal transducer and activator of transcription signal pathways, thereby indicating the immunoregulatory role of IL-35 in SLE. These in vivo findings may provide a biochemical basis for further investigation of the regulatory mechanisms of IL-35 for the treatment of autoimmune-mediated inflammation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV41764
Anti-GNAS (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV41765
Anti-GNAS (AB2) antibody produced in rabbit, IgG fraction of antiserum
AV42694
Anti-GNAS (AB3) antibody produced in rabbit, IgG fraction of antiserum
SAB2501411
Anti-GNAS antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA018122
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100941
Anti-GNAS antibody produced in rabbit, affinity isolated antibody
HPA027478
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA028386
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
SAB1400841
Anti-LIPH antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA049079
Anti-LIPH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107647
Anti-LIPH antibody produced in rabbit, affinity isolated antibody
HPA021656
Anti-SLC10A3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105728
Anti-SLC10A3 antibody produced in rabbit, affinity isolated antibody