EMAIL THIS PAGE TO A FRIEND
Genes & development
Oxygen-sensing PHDs regulate bone homeostasis through the modulation of osteoprotegerin.
PMID 25846796
Abstract
The bone microenvironment is composed of niches that house cells across variable oxygen tensions. However, the contribution of oxygen gradients in regulating bone and blood homeostasis remains unknown. Here, we generated mice with either single or combined genetic inactivation of the critical oxygen-sensing prolyl hydroxylase (PHD) enzymes (PHD1-3) in osteoprogenitors. Hypoxia-inducible factor (HIF) activation associated with Phd2 and Phd3 inactivation drove bone accumulation by modulating osteoblastic/osteoclastic cross-talk through the direct regulation of osteoprotegerin (OPG). In contrast, combined inactivation of Phd1, Phd2, and Phd3 resulted in extreme HIF signaling, leading to polycythemia and excessive bone accumulation by overstimulating angiogenic-osteogenic coupling. We also demonstrate that genetic ablation of Phd2 and Phd3 was sufficient to protect ovariectomized mice against bone loss without disrupting hematopoietic homeostasis. Importantly, we identify OPG as a HIF target gene capable of directing osteoblast-mediated osteoclastogenesis to regulate bone homeostasis. Here, we show that coordinated activation of specific PHD isoforms fine-tunes the osteoblastic response to hypoxia, thereby directing two important aspects of bone physiology: cross-talk between osteoblasts and osteoclasts and angiogenic-osteogenic coupling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV34608
Anti-EGLN1 antibody produced in rabbit, IgG fraction of antiserum
HPA022129
Anti-EGLN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV34483
Anti-EGLN2 antibody produced in rabbit, affinity isolated antibody
SAB2103176
Anti-EGLN2 antibody produced in rabbit, affinity isolated antibody
SAB2106483
Anti-EGLN2 antibody produced in rabbit, affinity isolated antibody
SAB2103253
Anti-EGLN2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2500342
Anti-EGLN3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1408202
Anti-EGLN3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV52402
Anti-EGLN3 antibody produced in rabbit, affinity isolated antibody
SAB1410173
Anti-EGLN3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA005928
Anti-EMCN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2700787
Anti-EMCN antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4502163
Anti-Mucin-14, C-Terminal antibody produced in rabbit, affinity isolated antibody
AV47546
Anti-UNCX antibody produced in rabbit, affinity isolated antibody
SAB2102654
Anti-UNCX antibody produced in rabbit, affinity isolated antibody
RAB0484
Human Osteoprotegerin / TNFRSF11b ELISA Kit, for serum, plasma, cell culture supernatant and urine
SAB1403432
Monoclonal Anti-EGLN2 antibody produced in mouse, clone 2G3, purified immunoglobulin, buffered aqueous solution
SAB1403433 Monoclonal Anti-EGLN3 antibody produced in mouse, clone 3C5, purified immunoglobulin, buffered aqueous solution
RAB0493
Mouse Osteoprotegerin ELISA Kit, for serum, plasma, cell culture supernatant
RAB0494
Mouse TRANCE ELISA Kit, for serum, plasma and cell culture supernatant
P3350000
Proline, European Pharmacopoeia (EP) Reference Standard
C5H9NO2