EMAIL THIS PAGE TO A FRIEND
Blood
Antagonism between MCL-1 and PUMA governs stem/progenitor cell survival during hematopoietic recovery from stress.
PMID 25847014
Abstract
Understanding the critical factors that govern recovery of the hematopoietic system from stress, such as during anticancer therapy and bone marrow transplantation, is of clinical significance. We investigated the importance of the prosurvival proteins myeloid cell leukemia-1 (MCL-1) and B-cell lymphoma-extra large (BCL-XL) in stem/progenitor cell survival and fitness during hematopoietic recovery from stress. Loss of a single Mcl-1 allele, which reduced MCL-1 protein levels, severely compromised hematopoietic recovery from myeloablative challenge and following bone marrow transplantation, whereas BCL-XL was dispensable in both contexts. We identified inhibition of proapoptotic p53 upregulated modulator of apoptosis (PUMA) as the key role of MCL-1 in both settings, with Mcl-1(+/-);Puma(-/-) mice completely protected from the deleterious effects of loss of 1 Mcl-1 allele. These results reveal the molecular mechanisms that govern cell survival during hematopoietic recovery from stress.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300489
Anti-KIT (Ab-721) antibody produced in rabbit, affinity isolated antibody
SAB4300593
Anti-KIT (Ab-936) antibody produced in rabbit, affinity isolated antibody
GW21114 Anti-KIT antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1412042
ANTI-KIT antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
SAB1412043
ANTI-KIT antibody produced in mouse, clone 5F6, purified immunoglobulin, buffered aqueous solution
SAB1412044
ANTI-KIT antibody produced in mouse, clone 5A11, purified immunoglobulin, buffered aqueous solution
SAB1412045
ANTI-KIT antibody produced in mouse, clone 2C3, purified immunoglobulin, buffered aqueous solution
SAB1412046
ANTI-KIT antibody produced in mouse, clone 1D3, purified immunoglobulin, buffered aqueous solution
SAB1412140
ANTI-KIT antibody produced in mouse, clone 4F7, purified immunoglobulin, buffered aqueous solution
SAB1412141
ANTI-KIT antibody produced in mouse, clone 3A8, purified immunoglobulin, buffered aqueous solution
HPA004471
Anti-KIT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501647
Anti-KIT, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4700750
Monoclonal Anti-CD117 antibody produced in mouse, clone 104D2, purified immunoglobulin, buffered aqueous solution
WH0003815M2
Monoclonal Anti-KIT antibody produced in mouse, clone 6F2, purified immunoglobulin, buffered aqueous solution
AMAB90901
Monoclonal Anti-KIT antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1657, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90904
Monoclonal Anti-KIT antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1667, purified immunoglobulin, buffered aqueous glycerol solution
SAB1404007
Monoclonal Anti-KIT antibody produced in mouse, clone 6G12, purified immunoglobulin, buffered aqueous solution
SAB5300439
Monoclonal Anti-KIT antibody produced in mouse, clone 8D7, ascites fluid
SAB5300440
Monoclonal Anti-KIT antibody produced in mouse, clone 1C5, ascites fluid