EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Diverse effects of LPA4, LPA5 and LPA6 on the activation of tumor progression in pancreatic cancer cells.
PMID 25849892
Abstract
Lysophosphatidic acid (LPA) is an extracellular biological lipid which interacts with G protein-coupled LPA receptors (LPA1 to LPA6). LPA signaling via LPA receptors mediates several cellular responses. In the present study, to assess the roles of LPA4, LPA5 and LPA6 in cellular functions of pancreatic cancer cells, we generated LPA receptor knockdown cells from PANC-1 cells (PANC-sh4, PANC-sh5 and PANC-sh6 cells, respectively). In cell motility assay, PANC-sh4 and PANC-sh5 cells enhanced the cell motile activities, compared with control cells. In contrast, the cell motile activity of PANC-sh6 cells was suppressed. The invasive activities of PANC-sh4 and PANC-sh5 cells were markedly stimulated, while PANC-sh6 cells showed the low invasive activity. In colony assay, PANC-sh4 and PANC-sh5 cells formed the large sized colonies, but not PANC-sh6 cells. When endothelial cells were incubated with supernatants from PANC-sh4 and PANC-sh5 cells, the cell motility and tube formation of endothelial cells were significantly induced, but not PANC-sh6 cells. These results suggest that the diverse roles of LPA4, LPA5 and LPA6 are involved in the activation of tumor progression in pancreatic cancer cells.