EMAIL THIS PAGE TO A FRIEND
Nature communications
A cohesin-OCT4 complex mediates Sox enhancers to prime an early embryonic lineage.
PMID 25851587
Abstract
Short- and long-scales intra- and inter-chromosomal interactions are linked to gene transcription, but the molecular events underlying these structures and how they affect cell fate decision during embryonic development are poorly understood. One of the first embryonic cell fate decisions (that is, mesendoderm determination) is driven by the POU factor OCT4, acting in concert with the high-mobility group genes Sox-2 and Sox-17. Here we report a chromatin-remodelling mechanism and enhancer function that mediate cell fate switching. OCT4 alters the higher-order chromatin structure at both Sox-2 and Sox-17 loci. OCT4 titrates out cohesin and switches the Sox-17 enhancer from a locked (within an inter-chromosomal Sox-2 enhancer/CCCTC-binding factor CTCF/cohesin loop) to an active (within an intra-chromosomal Sox-17 promoter/enhancer/cohesin loop) state. SALL4 concomitantly mobilizes the polycomb complexes at the Soxs loci. Thus, OCT4/SALL4-driven cohesin- and polycombs-mediated changes in higher-order chromatin structure mediate instruction of early cell fate in embryonic cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4200034
Anti-BMI1 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution
SAB4200033
Anti-BMI1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA030472
Anti-BMI1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106309
Anti-BMI1 antibody produced in rabbit, affinity isolated antibody
SAB2501524 Anti-NIPBL antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA040834
Anti-NIPBL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV34213
Anti-PCGF4 antibody produced in rabbit, affinity isolated antibody
SAB2501454
Anti-POU5F1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV100956
Anti-POU5F1 antibody produced in rabbit, IgG fraction of antiserum
SAB2101850
Anti-POU5F1 antibody produced in rabbit, affinity isolated antibody
SAB3500316
Anti-POU5F1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701958
Anti-POU5F1 antibody produced in rabbit
SAB2701960
Anti-POU5F1 antibody produced in rabbit
SAB2701972
Anti-POU5F1 antibody produced in rabbit
HPA020044
Anti-RAD21 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107208
Anti-RAD21 antibody produced in rabbit, affinity isolated antibody
SAB2107209
Anti-RAD21 antibody produced in rabbit, affinity isolated antibody
SAB4502562
Anti-RAD21, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2500906
Anti-SALL4 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA015291
Anti-SALL4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA015791
Anti-SALL4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2702041
Anti-SALL4 antibody produced in rabbit
AV39190
Anti-SUZ12 (AB2) antibody produced in rabbit, affinity isolated antibody
HPA057436
Anti-SUZ12 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3501103
Anti-SUZ12 antibody produced in rabbit, affinity isolated antibody
EHU037061 MISSION® esiRNA, esiRNA human SALL4 (esiRNA1)
EHU083971 MISSION® esiRNA, esiRNA human NIPBL (esiRNA1)
EMU045581 MISSION® esiRNA, esiRNA targeting mouse Nipbl (esiRNA1)
EMU080881 MISSION® esiRNA, esiRNA targeting mouse Sall4 (esiRNA1)
SAB5300333
Monoclonal Anti-BMI1 antibody produced in mouse, clone 3E3, ascites fluid