EMAIL THIS PAGE TO A FRIEND
American journal of epidemiology
Obesity and Prostate Cancer Risk According to Tumor TMPRSS2:ERG Gene Fusion Status.
PMID 25852077
Abstract
The T2E gene fusion, formed by fusion of the transmembrane protease, serine 2, gene (TMPRSS2) with the erythroblast transformation-specific (ETS)-related gene (ERG), is found in approximately 50% of prostate cancers and may characterize distinct molecular subtypes of prostate cancer with different etiologies. We investigated the relationship between body mass index (BMI; weight (kg)/height (m)(2)) and prostate cancer risk by T2E status. Study participants were residents of King County, Washington, recruited for 2 population-based case-control studies conducted in 1993-1996 and 2002-2005. Tumor T2E status was determined for 563 prostate cancer patients who underwent radical prostatectomy. Information on weight, height, and covariables was obtained through in-person interviews. We performed polytomous logistic regression to calculate odds ratios and 95% confidence intervals for T2E-positive and -negative prostate cancer. Comparing the highest BMI quartile with the lowest, inverse associations were observed between recent (≥29.7 vs. <24.5: odds ratio = 0.66, 95% confidence interval: 0.45, 0.97) and maximum (≥31.8 vs. <25.9: odds ratio = 0.69, 95% confidence interval: 0.47, 1.02) BMI and the risk of T2E-positive prostate cancer. No significant associations were seen for men with T2E-negative tumors. This study provides evidence that obesity is specifically associated with reduced risk of developing androgen-responsive T2E fusion-positive tumors. The altered steroid hormone profile in obese men may contribute to this inverse association.