EMAIL THIS PAGE TO A FRIEND
Cardiovascular research
DNA-dependent protein kinase (DNA-PK) permits vascular smooth muscle cell proliferation through phosphorylation of the orphan nuclear receptor NOR1.
PMID 25852083
Abstract
Being central part of the DNA repair machinery, DNA-dependent protein kinase (DNA-PK) seems to be involved in other signalling processes, as well. NOR1 is a member of the NR4A subfamily of nuclear receptors, which plays a central role in vascular smooth muscle cell (SMC) proliferation and in vascular proliferative processes. We determined putative phosphorylation sites of NDA-PK in NOR1 and hypothesized that the enzyme is able to modulate NOR1 signalling and, this way, proliferation of SMC. Cultured human aortic SMC were treated with the specific DNA-PK inhibitor NU7026 (or siRNA), which resulted in a 70% inhibition of FCS-induced proliferation as measured by BrdU incorporation. Furthermore, FCS-stimulated up-regulation of NOR1 protein as well as the cell-cycle promoting proteins proliferating cell nuclear antigen (PCNA), cyclin D1, and hyperphosphorylation of the retinoblastoma protein were prevented by DNA-PK inhibition. Co-immunoprecipitation studies from VSM cell lysates demonstrated that DNA-PK forms a complex with NOR1. Mutational analysis and kinase assays demonstrated that NOR1 is a substrate of DNA-PK and is phosphorylated in the N-terminal domain. Phosphorylation resulted in post-transcriptional stabilization of the protein through prevention of its ubiquitination. Active DNA-PK and NOR1 were found predominantly expressed within the neointima of human atherosclerotic tissue specimens. In mice, inhibition of DNA-PK significantly attenuated neointimal lesion size 3 weeks after wire-injury. DNA-PK directly phosphorylates NOR-1 and, this way, modulates SMC proliferation. These data add to our understanding of vascular remodelling processes and opens new avenues for treatment of vascular proliferative diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1537
NU7026, ≥98% (HPLC), solid
C17H15NO3