EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
A cytosolic heat shock protein 90 and cochaperone CDC37 complex is required for RIP3 activation during necroptosis.
PMID 25852146
Abstract
Receptor-interacting protein kinase 3, RIP3, and a pseudokinase mixed lineage kinase-domain like protein, MLKL, constitute the core components of the necroptosis pathway, which causes programmed necrotic death in mammalian cells. Latent RIP3 in the cytosol is activated by several upstream signals including the related kinase RIP1, which transduces signals from the tumor necrosis factor (TNF) family of cytokines. We report here that RIP3 activation following the induction of necroptosis requires the activity of an HSP90 and CDC37 cochaperone complex. This complex physically associates with RIP3. Chemical inhibitors of HSP90 efficiently block necroptosis by preventing RIP3 activation. Cells with knocked down CDC37 were unable to respond to necroptosis stimuli. Moreover, an HSP90 inhibitor that is currently under clinical development as a cancer therapy was able to prevent systemic inflammatory response syndrome in rats treated with TNF-α. HSP90 and CDC37 cochaperone complex-mediated protein folding is thus an important part of the RIP3 activation process during necroptosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA003928
Anti-CDC37 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
M6697
Anti-MLKL (58-70) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1408428
Anti-MLKL antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2103620
Anti-MLKL, (N-terminal) antibody produced in rabbit, affinity isolated antibody
HPA046651
Anti-RALBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410141
Anti-RALBP1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2105823
Anti-RALBP1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
GW21168 Anti-RIPK1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA015257
Anti-RIPK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV02038
Anti-RIPK1 antibody produced in rabbit, affinity isolated antibody
SAB3500420
Anti-RIPK1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1401396
Anti-UQCRFS1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401397
Anti-UQCRFS1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA041863
Anti-UQCRFS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105078
Anti-UQCRFS1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
WH0011140M1
Monoclonal Anti-CDC37 antibody produced in mouse, clone 3C7, purified immunoglobulin, buffered aqueous solution
SAB1402671
Monoclonal Anti-MLKL, (C-terminal) antibody produced in mouse, clone 3B2, purified immunoglobulin, buffered aqueous solution
WH0010928M2
Monoclonal Anti-RALBP1 antibody produced in mouse, clone 2A1, purified immunoglobulin, buffered aqueous solution
SAB1404880 Monoclonal Anti-RALBP1 antibody produced in mouse, clone 1C2-A10, purified immunoglobulin, buffered aqueous solution
SAB4100176
Monoclonal Anti-RALBP1 antibody produced in mouse, clone RALBP1-A192, culture supernatant
71726
Sodium dodecyl sulfate, for ion pair chromatography, ≥99.0%
C12H25NaO4S
71727
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
C12H25NaO4S
Y0000620
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
C12H25NaO4S