EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Cell physiology
The Golgi apparatus regulates cGMP-dependent protein kinase I compartmentation and proteolysis.
PMID 25855081
Abstract
cGMP-dependent protein kinase I (PKGI) is an important effector of cGMP signaling that regulates vascular smooth muscle cell (SMC) phenotype and proliferation. PKGI has been detected in the perinuclear region of cells, and recent data indicate that proprotein convertases (PCs) typically resident in the Golgi apparatus (GA) can stimulate PKGI proteolysis and generate a kinase fragment that localizes to the nucleus and regulates gene expression. However, the role of the endomembrane system in PKGI compartmentation and processing is unknown. Here, we demonstrate that PKGI colocalizes with endoplasmic reticulum (ER), ER-Golgi intermediate compartment, GA cisterna, and trans-Golgi network proteins in pulmonary artery SMC and cell lines. Moreover, PKGI localizes with furin, a trans-Golgi network-resident PC known to cleave PKGI. ER protein transport influences PKGI localization because overexpression of a constitutively inactive Sar1 transgene caused PKGI retention in the ER. Additionally, PKGI appears to reside within the GA because PKGI immunoreactivity was determined to be resistant to cytosolic proteinase K treatment in live cells. The GA appears to play a role in PKGI proteolysis because overexpression of inositol 1,4,5-trisphosphate receptor-associated cGMP kinase substrate, not only tethered heterologous PKGI-β to the ER and decreased its localization to the GA, but also diminished PKGI proteolysis and nuclear translocation. Also, inhibiting intra-GA protein transport with monensin was observed to decrease PKGI cleavage. These studies detail a role for the endomembrane system in regulating PKGI compartmentation and proteolysis. Moreover, they support the investigation of mechanisms regulating PKGI-dependent nuclear cGMP signaling in the pulmonary vasculature with Golgi dysfunction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1303143
ANTI-EREG (C-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1408982
Anti-EREG antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
E1031
Anti-ERGIC-53/p58 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300344
Anti-ESR1 (Ab-104) antibody produced in rabbit, affinity isolated antibody
SAB4300345
Anti-ESR1 (Ab-106) antibody produced in rabbit, affinity isolated antibody
SAB4300346
Anti-ESR1 (Ab-118) antibody produced in rabbit, affinity isolated antibody
SAB4300347
Anti-ESR1 (Ab-167) antibody produced in rabbit, affinity isolated antibody
SAB2100711
Anti-ESR1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100712
Anti-ESR1 (ab2) antibody produced in rabbit, affinity isolated antibody
GW21182 Anti-ESR1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA000449
Anti-ESR1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA000450
Anti-ESR1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104971
Anti-ESR1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
E1396 Anti-Estrogen Receptor (154-171) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
E0521 Anti-Estrogen Receptor (18-32) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
G7295
Anti-GM130 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1405886
Anti-GOLGA2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA021178
Anti-GOLGA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA021230
Anti-GOLGA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA021799
Anti-GOLGA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3
AV48150
Anti-P4HB (AB1) antibody produced in rabbit, affinity isolated antibody
AV48151
Anti-P4HB (AB2) antibody produced in rabbit, affinity isolated antibody
SAB1406213
Anti-P4HB antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA018884
Anti-P4HB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
P7122
Anti-Protein Disulfide Isomerase (DL-11) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
P7372
Anti-Protein Disulfide Isomerase (MD-12) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
P7496
Anti-Protein Disulfide Isomerase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
T9826
Anti-TGN38 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
T7576
Anti-TGN46 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA012609
Anti-TGOLN2 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution