EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Egr3-dependent muscle spindle stretch receptor intrafusal muscle fiber differentiation and fusimotor innervation homeostasis.
PMID 25855173
Abstract
Muscle stretch proprioceptors (muscle spindles) are required for stretch reflexes and locomotor control. Proprioception abnormalities are observed in many human neuropathies, but the mechanisms involved in establishing and maintaining muscle spindle innervation and function are still poorly understood. During skeletal muscle development, sensory (Ia-afferent) innervation induces contacted myotubes to transform into intrafusal muscle fibers that form the stretch receptor core. The transcriptional regulator Egr3 is induced in Ia-afferent contacted myotubes by Neuregulin1 (Nrg1)/ErbB receptor signaling and it has an essential role in spindle morphogenesis and function. Because Egr3 is widely expressed during development and has a pleiotropic function, whether Egr3 functions primarily in skeletal muscle, Ia-afferent neurons, or in Schwann cells that myelinate Ia-afferent axons remains unresolved. In the present studies, cell-specific ablation of Egr3 in mice showed that it has a skeletal muscle autonomous function in stretch receptor development. Moreover, using genetic tracing, we found that Ia-afferent contacted Egr3-deficient myotubes were induced in normal numbers, but their development was blocked to generate one to two shortened fibers that failed to express some characteristic myosin heavy chain (MyHC) proteins. These "spindle remnants" persisted into adulthood, remained innervated by Ia-afferents, and expressed neurotrophin3 (NT3), which is required for Ia-afferent neuron survival. However, they were not innervated by fusimotor axons and they did not express glial derived neurotrophic factor (GDNF), which is essential for fusimotor neuron survival. These results demonstrate that Egr3 has an essential role in regulating gene expression that promotes normal intrafusal muscle fiber differentiation and fusimotor innervation homeostasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56671
(±)-Propylene oxide, analytical standard
C3H6O
HPA006206
Anti-EGR3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104196
Anti-EGR3 antibody produced in rabbit, affinity isolated antibody
SAB2104801
Anti-EGR3 antibody produced in rabbit, affinity isolated antibody
SAB2105230
Anti-EGR3 antibody produced in rabbit, affinity isolated antibody
SAB2104663
Anti-EGR3, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4500726
Anti-EGR3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1401150
Anti-GDNF antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1300906
Anti-NTF3 (C-term) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1300907
Anti-NTF3 (center) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1400188
Anti-NTF3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA032000
Anti-NTF3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA006889
Anti-SORT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
S0697
Anti-Sortilin antibody produced in rabbit, lyophilized powder, affinity isolated antibody
T2450 Anti-TrkC antibody produced in goat, affinity isolated antibody
G1128
Tragacanth