EMAIL THIS PAGE TO A FRIEND
Parasitology research
Exosome-like vesicles derived by Schistosoma japonicum adult worms mediates M1 type immune- activity of macrophage.
PMID 25855345
Abstract
Exosomes are 30-100-nm membrane vesicles of endocytic origin that are released into the extracellular space upon fusion of the multi-vesicular bodies (MVB) with the plasma membrane, while initial studies described that the role of exosomes was a reticulocyte cargo-disposal mechanism allowing remodeling of the plasma membrane during the maturation of reticulocytes to erythrocytes. Recent studies indicate that exosomes are secreted by most cells and pathogens and play an important role in intercellular signaling and exert regulatory function by carrying bioactive molecules. As numerous pathogens, adult worm of Schistosoma japonicum (S. japonicum) reside in mesenteric veins of definitive host including man and mammal animals. It was reported that the worms or the eggs also have specialized secretion systems to export effector proteins or other molecules into host target cells. However, the mechanisms involved remained unclear. This study investigated the isolation of the exosome-like vesicles secreted by S. japonicum adult worms and its immune activity on microphage in vitro. In this report, we identified exosome-based secretion as a new mechanism for protein secretion by S. japonicum. Electron microscopy tomography revealed the previously unidentified ultrastructural detail of exosome-like vesicles with high resolution; they were found to be typical spherical shape and to have a diverse population that varies in size of 30-100xa0nm. Exosome-like vesicles isolated from S. japonicum contained a significantly different protein compared with debris pelleted and the apoptosis body. We also demonstrate that macrophages were preferentially differentiated into the M1 subtype while being treated with S. japonicum exosome-like vesicles. This study reveals there are exosome-like vesicles derived by S. japonicum adult worms, and the exosome-like vesicles can mediate M1-type immune- activity of macrophage.