PloS one
RAP1 GTPase overexpression is associated with cervical intraepithelial neoplasia.
PMID 25856570
Abstract
RAP1 (RAS proximate 1), a small GTP-binding protein of the RAS superfamily, is a putative oncogene that is highly expressed in several malignant cell lines and types of cancers, including some types of squamous cell carcinoma. However, the participation of RAP1 in cervical carcinogenesis is unknown. We conducted a cross-sectional study of paraffin-embedded cervical biopsies to determine the association of RAP1 with cervical intraepithelial neoplasia (CIN). Standard and quantitative immunohistochemistry assessment of RAP1 expression in fixed tissue was performed on 183 paraffin-embedded cervical biopsies that were classified as normal or non-dysplastic mucosa (NDM) (n = 33); CIN grade 1 (n = 84) and CIN grade 2/3 (n = 66). A gradual increase in RAP1 expression in NDM < CIN 1 < CIN 2/3 (p<0.001) specimens was observed and was in agreement with the histopathologic diagnosis. A progressive increase in the RAP1 expression levels increased the risk of CIN 1 [odds ratio (OR) = 3.50; 95% confidence interval (CI) 1.30-10.64] 3.5 fold and the risk of CIN 2/3 (OR = 19.86, 95% CI 6.40-70.79) nearly 20 fold when compared to NDM. In addition, stereotype ordinal regression analysis showed that this progressive increase in RAP1 expression more strongly impacted CIN 2/3 than CIN 1. Our findings suggest that RAP1 may be a useful biomarker for the diagnosis of CIN.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R5405
Anti-Rabex-5 (N-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB3500995
Anti-RABEX5 antibody produced in rabbit, affinity isolated antibody
SAB1407359
Anti-RABGEF1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2700823
Anti-RAP1A antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2501551
Anti-TERF2IP (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA006719
Anti-TERF2IP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106088
Anti-TERF2IP antibody produced in rabbit, affinity isolated antibody
SAB2107262
Anti-TERF2IP antibody produced in rabbit, affinity isolated antibody
R8154 Monoclonal Anti-RAP1 antibody produced in mouse, ~2 mg/mL, clone 4C8/1, purified immunoglobulin, buffered aqueous solution
WH0005906M1
Monoclonal Anti-RAP1A antibody produced in mouse, clone 3E1, purified immunoglobulin, buffered aqueous solution
SAB5300214
Monoclonal Anti-RAP1A antibody produced in mouse, clone 5F8, purified immunoglobulin, buffered aqueous solution
SAB5300215
Monoclonal Anti-RAP1A antibody produced in mouse, clone 5F8G2, purified immunoglobulin, buffered aqueous solution