EMAIL THIS PAGE TO A FRIEND
Phytotherapy research : PTR
Korean Red Ginseng Extract Enhances the Anticancer Effects of Imatinib Mesylate Through Abrogation p38 and STAT5 Activation in KBM-5 Cells.
PMID 25857479
Abstract
Although imatinib mesylate (IM) in the treatment of chronic myelogenous leukemia (CML) remains the best example of successful targeted therapy, majority of patients with CML suffer its toxicity profile and develop chemoresistance to existing therapeutic agents. Thus, there is a need to develop novel alternative therapies for the treatment of CML. Here, we investigated whether Korean red ginseng extract (KRGE) could suppress the proliferation and induce chemosensitization in human CML cells. Also, we used a human phospho-antibody array containing 46 antibodies against signaling molecules to examine a subset of phosphorylation events after treatment. Korean red ginseng extract broadly suppressed the proliferation of five different cell lines, but KRGE was found to be the most potent inducer of apoptosis against KBM-5 cells. It also abrogated the expression of Bcl-2 (B-cell lymphoma 2), Bcl-xL (B-cell lymphoma-extra large), survivin, inhibitors of apoptosis protein 1/2, COX-2 (Cyclooxygenase-2), cyclin D1, matrix metalloproteinase-9, and VEGF (Vascular endothelial growth factor), as well as upregulated the expression of pro-apoptotic gene products. Interestingly, KRGE also enhanced the cytotoxic and apoptotic effect of IM in KBM-5 cells. The combination treatment of KRGE and IM caused pronounced suppression of p38 and signal transducer and activator of transcription 5 phosphorylation and induced phosphorylation of p53 compared with the individual treatment. Our results demonstrate that KRGE can enhance the anticancer activity of IM and may have a substantial potential in the treatment of CML.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA005513
Anti-BIRC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401012
Anti-BIRC2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501559
Anti-BIRC3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501560
Anti-BIRC3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1409216
Anti-BIRC3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002317
Anti-BIRC3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401013
Anti-BIRC3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4503514
Anti-Cyclin E1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4504499
Anti-phospho-p53 (pSer15) antibody produced in rabbit, affinity isolated antibody
SAB4503953
Anti-phospho-p53 (pSer20) antibody produced in rabbit, affinity isolated antibody
SAB4504500
Anti-phospho-p53 (pSer315) antibody produced in rabbit, affinity isolated antibody
SAB4504501
Anti-phospho-p53 (pSer33) antibody produced in rabbit, affinity isolated antibody
SAB4504592
Anti-phospho-p53 (pSer366) antibody produced in rabbit, affinity isolated antibody
SAB4504502
Anti-phospho-p53 (pSer37) antibody produced in rabbit, affinity isolated antibody
SAB4504593
Anti-phospho-p53 (pSer378) antibody produced in rabbit, affinity isolated antibody
P8982 Anti-phospho-p53 (pSer392) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503954
Anti-phospho-p53 (pSer392) antibody produced in rabbit, affinity isolated antibody
SAB4504503
Anti-phospho-p53 (pSer46) antibody produced in rabbit, affinity isolated antibody
SAB4504504
Anti-phospho-p53 (pSer6) antibody produced in rabbit, affinity isolated antibody
SAB4504505
Anti-phospho-p53 (pSer9) antibody produced in rabbit, affinity isolated antibody
SAB4504506
Anti-phospho-p53 (pThr18) antibody produced in rabbit, affinity isolated antibody
SAB4504591
Anti-phospho-p53 (pThr387) antibody produced in rabbit, affinity isolated antibody
SAB1306688 ANTI-PHOSPHO-P53(S20) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305629 ANTI-PHOSPHO-P53(S315) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306606
ANTI-PHOSPHO-P53(S9) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306609 ANTI-PHOSPHO-P53(T18) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305574 ANTI-PHOSPHO-P53(T55) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300073
Anti-phospho-TP53 (pSer15) antibody produced in rabbit, affinity isolated antibody
SAB4300079
Anti-phospho-TP53 (pSer315) antibody produced in rabbit, affinity isolated antibody
SAB4300076
Anti-phospho-TP53 (pSer33) antibody produced in rabbit, affinity isolated antibody