EMAIL THIS PAGE TO A FRIEND
Alimentary pharmacology & therapeutics
Cultured blood T-cell responses predict anti-TNF therapy response in patients with ulcerative colitis.
PMID 25858346
Abstract
Anti-tumour necrosis factor (TNF) therapy is used for treatment of ulcerative colitis (UC). As approximately 30% of patients with UC do not benefit from the treatment, it is of clinical interest to identify biomarkers of response before therapy is initiated. To identify prognostic biomarkers of anti-TNF therapy response in anti-TNF therapy-naïve patients with UC. Peripheral blood cells were obtained from 56 patients with UC before therapy started. Thirty-four patients were included in an exploratory cohort and 22 patients in a validation cohort. Blood cells were stimulated in vitro with influenza vaccine with and without anti-TNF. T-cell surface receptor expression and cytokine release were determined (in total 17 variables). Treatment response was evaluated using the Mayo score 12-14xa0weeks after the first infusion. In the exploratory cohort, blood cells from the patients showed stronger anti-TNF-dependent suppression of T-cell surface receptor expression and cytokine secretion among therapy responders than nonresponders. In particular, anti-TNF suppressed the expression of CD25 on T cells and secretion of interleukin 5, to a higher degree in responders than in nonresponders. These variables were used to a create model to predict therapy outcome, which was confirmed in the validation cohort. Correct classification of future therapy response was achieved in 91% of the cases in the validation cohort. The effects of anti-TNF on cultured blood T cells, obtained before therapy started, predict treatment outcome in patients with UC.