EMAIL THIS PAGE TO A FRIEND
Nature communications
Regulation of autophagy and the ubiquitin-proteasome system by the FoxO transcriptional network during muscle atrophy.
PMID 25858807
Abstract
Stresses like low nutrients, systemic inflammation, cancer or infections provoke a catabolic state characterized by enhanced muscle proteolysis and amino acid release to sustain liver gluconeogenesis and tissue protein synthesis. These conditions activate the family of Forkhead Box (Fox) O transcription factors. Here we report that muscle-specific deletion of FoxO members protects from muscle loss as a result of the role of FoxOs in the induction of autophagy-lysosome and ubiquitin-proteasome systems. Notably, in the setting of low nutrient signalling, we demonstrate that FoxOs are required for Akt activity but not for mTOR signalling. FoxOs control several stress-response pathways such as the unfolded protein response, ROS detoxification, DNA repair and translation. Finally, we identify FoxO-dependent ubiquitin ligases including MUSA1 and a previously uncharacterised ligase termed SMART (Specific of Muscle Atrophy and Regulated by Transcription). Our findings underscore the central function of FoxOs in coordinating a variety of stress-response genes during catabolic conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1306343
ANTI-FKHR (CENTER D469) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
F2303 Anti-FKHR (FOXO1a) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500938
Anti-FKHR antibody produced in rabbit, affinity isolated antibody
SAB4500939
Anti-FKHR antibody produced in rabbit, affinity isolated antibody
F2178 Anti-FKHRL1 (FOXO3a) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500941
Anti-FKHRL1 antibody produced in rabbit, affinity isolated antibody
SAB4300410
Anti-FOXO1 (Ab-256) antibody produced in rabbit, affinity isolated antibody
SAB4300428
Anti-FOXO1 (Ab-319) antibody produced in rabbit, affinity isolated antibody
AV100607
Anti-FOXO1 (AB1) antibody produced in rabbit, affinity isolated antibody
AV32107
Anti-FOXO1 (AB2) antibody produced in rabbit, affinity isolated antibody
AV32976
Anti-FOXO1 (AB3) antibody produced in rabbit, affinity isolated antibody
SAB2500417
Anti-FOXO1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA001252
Anti-FOXO1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500507
Anti-FOXO1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1411790
ANTI-FOXO1A antibody produced in mouse, clone 4B4, purified immunoglobulin, buffered aqueous solution
SAB1411812
ANTI-FOXO1A antibody produced in mouse, clone 3H5, purified immunoglobulin, buffered aqueous solution
SAB1411813
ANTI-FOXO1A antibody produced in mouse, clone 4A3, purified immunoglobulin, buffered aqueous solution
SAB1411814
ANTI-FOXO1A antibody produced in mouse, clone 4A10, purified immunoglobulin, buffered aqueous solution
SAB1411815
ANTI-FOXO1A antibody produced in mouse, clone 2F12, purified immunoglobulin, buffered aqueous solution
SAB1411857
ANTI-FOXO1A antibody produced in mouse, clone 1F11, purified immunoglobulin, buffered aqueous solution
SAB1411858
ANTI-FOXO1A antibody produced in mouse, clone 4A2, purified immunoglobulin, buffered aqueous solution
SAB1411859
ANTI-FOXO1A antibody produced in mouse, clone 4D9, purified immunoglobulin, buffered aqueous solution
SAB1411860
ANTI-FOXO1A antibody produced in mouse, clone 1A6, purified immunoglobulin, buffered aqueous solution
SAB1411861
ANTI-FOXO1A antibody produced in mouse, clone 1F6, purified immunoglobulin, buffered aqueous solution
AV38094
Anti-FOXO1A antibody produced in rabbit, IgG fraction of antiserum
SAB4500936
Anti-FOXO1A antibody produced in rabbit, affinity isolated antibody
SAB4500937
Anti-FOXO1A antibody produced in rabbit, affinity isolated antibody
AV38040
Anti-FOXO3 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3500508
Anti-FOXO3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1411796
ANTI-FOXO3A antibody produced in mouse, clone 3F12, purified immunoglobulin, buffered aqueous solution