EMAIL THIS PAGE TO A FRIEND
Acta histochemica et cytochemica
Changes in laminin chain expression in pre- and postnatal rat pituitary gland.
PMID 25861129
Abstract
Cell-matrix interaction is required for tissue development. Laminin, a major constituent of the basement membrane, is important for structural support and as a ligand in tissue development. Laminin has 19 isoforms, which are determined by combinational assembly of five α, three β, and three γ chains (eg, laminin 121 is α1, β2, and γ1). However, no report has identified the laminin isoforms expressed during pituitary development. We used in situ hybridization to investigate all laminin chains expressed during rat anterior pituitary development. The α5 chain was expressed during early pituitary development (embryonic day 12.5-15.5). Expression of α1 and α4 chains was noted in vasculature cells at embryonic day 19.5, but later diminished. The α1 chain was re-expressed in parenchymal cells of anterior lobe from postnatal day 10 (P10), while the α4 chain was present in vasculature cells from P30. The α2 and α3 chains were transiently expressed in vasculature cells and anterior lobe, respectively, only at P30. Widespread distribution of β and γ chains was also observed during development. These findings suggest that numerous laminin isoforms are involved in anterior pituitary gland development and that alteration of the expression pattern is required for proper development of the gland.