EMAIL THIS PAGE TO A FRIEND
Acta neuropathologica
Identification of a novel MET mutation in high-grade glioma resulting in an auto-active intracellular protein.
PMID 25862637
Abstract
MET has gained interest as a therapeutic target for a number of malignancies because of its involvement in tumorigenesis, invasion and metastasis. At present, a number of inhibitors, both antibodies against MET or its ligand hepatocyte growth factor, and small molecule MET tyrosine kinase inhibitors are in clinical trials. We here describe a novel variant of MET that is expressed in 6% of high-grade gliomas. Characterization of this mutation in a glioma cell line revealed that it consists of an intronic deletion, resulting in a splice event connecting an intact splice donor site in exon 6 with the next splice acceptor site being that of exon 9. The encoded protein lacks parts of the extracellular IPT domains 1 and 2, encoded by exons 7 and 8, resulting in a novel pseudo-IPT and is named MET(Δ7-8). MET(Δ7-8) is located predominantly in the cytosol and is constitutively active. The auto-activating nature of MET(Δ7-8), in combination with a lack of transmembrane localization, renders MET(Δ7-8) not targetable using antibodies, although the protein is efficiently deactivated by MET-specific tyrosine kinase inhibitors. Testing of MET-expressing tumors for the presence of this variant may be important for treatment decision making.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H9786 Anti-Hepatocyte Growth Factor Receptor (c-Met) antibody produced in goat, affinity isolated antibody, lyophilized powder
H9911 Anti-Hepatocyte Growth Factor Receptor antibody produced in goat, lyophilized powder, affinity isolated antibody
SAB4300599
Anti-MET (Ab-1003) antibody produced in rabbit, affinity isolated antibody
SAB4300479
Anti-MET (Ab-1234) antibody produced in rabbit, affinity isolated antibody
SAB4300487
Anti-MET (Ab-1349) antibody produced in rabbit, affinity isolated antibody
SAB1406117
Anti-MET antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4501870
Anti-MET antibody produced in rabbit, affinity isolated antibody
SAB4501868
Anti-MET, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501869
Anti-MET, C-Terminal antibody produced in rabbit, affinity isolated antibody
C7240 Anti-phospho-c-Met (pTyr1230/pTyr1234/pTyr1235) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
C6990 Anti-phospho-c-Met (pTyr1349) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300182
Anti-phospho-MET (pTyr1234) antibody produced in rabbit, affinity isolated antibody
SAB4300190
Anti-phospho-MET (pTyr1349) antibody produced in rabbit, affinity isolated antibody
AV40493
Anti-RNMT antibody produced in rabbit, affinity isolated antibody
SAB2102036
Anti-RNMT antibody produced in rabbit, affinity isolated antibody
HPA039409
Anti-RNMT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0008731M1
Monoclonal Anti-RNMT antibody produced in mouse, clone 3H3-1D12, purified immunoglobulin, buffered aqueous solution