EMAIL THIS PAGE TO A FRIEND
World neurosurgery
Meningioma in Down Syndrome.
PMID 25862935
Abstract
Down syndrome comprises multiple malformations and is due to trisomy of chromosome 21. There is epidemiologic evidence that individuals with Down syndrome are at decreased risk for solid tumors including brain tumors. It has been suggested that some genes expressed on the extra copy of chromosome 21 act as tumor suppressor genes and contribute to protection against tumorigenesis. We report the first case to our knowledge of a patient with Down syndrome, an 8-year-old boy, with an intracranial meningioma, in which the status of chromosome 21 was examined. The diagnosis was based on histologic examination of the surgically resected tumor. Postoperatively, the patient's neurologic status improved, and there was no tumor regrowth in the next 2 years. Fluorescence in situ hybridization for chromosome 22 confirmed high allele loss involving the neurofibromin 2 gene locus, a finding typical in meningiomas. Fluorescence in situ hybridization also revealed chromosome 21 heterogeneity in tumor cells; not only cells with trisomy 21 but also cells with disomy and monosomy 21 were present. All blood cells from the patient manifested trisomy 21. Deletion of the chromosome 21 allele may be associated with tumorigenesis of meningioma in Down syndrome. This supports the hypothesis that some genes whose expression is increased on the extra copy of chromosome 21 function as tumor suppressor genes and that they contribute to the reduced tumor incidence in individuals with Down syndrome.