EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Monitoring tryptophan metabolism after exposure to hexaconazole and the enantioselective metabolism of hexaconazole in rat hepatocytes in vitro.
PMID 25863579
Abstract
In the present study, the enantioselective metabolism, cytotoxicity of hexaconazole and its influence on tryptophan metabolism in rat hepatocytes in vitro were investigated. Following the exposure of primary rat hepatocytes to rac-hexaconazole, the concentrations of its enantiomers in the media were determined by chiral high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS). The half-lives (t1/2) of (+)-hexaconazole and (-)-hexaconazole were 5.17 h and 19.80 h, respectively, indicating that the metabolic process was enantioselective with (-)-hexaconazole enrichment. Using the MTT method, the EC50 values of rac-hexaconazole, (+)-hexaconazole and (-)-hexaconazole after 12h of exposure were determined to be 71.62, 62.71 and 67.94 μM, respectively. Tryptophan metabolism was monitored using metabolomics profiling techniques. Hexaconazole and its enantiomers caused the down-regulation of tryptophan levels and the up-regulation of kynurenine (KYN) levels, suggesting a role for hexaconazole in the activation of the KYN pathway and providing information for the mechanism of its toxicity.