EMAIL THIS PAGE TO A FRIEND
Virus research
The heterogeneous nuclear ribonucleoprotein K (hnRNP K) is a host factor required for dengue virus and Junín virus multiplication.
PMID 25865411
Abstract
Heterogeneous nuclear ribonucleoproteins (hnRNPs) are cellular factors involved in the replication of several viruses. In this study we analyzed the expression and intracellular localization of hnRNP A2 and hnRNP K in cell cultures infected with two viruses that cause human hemorrhagic fevers: dengue virus type 2 (DENV-2) and Junín virus (JUNV). We determined that DENV-2 promoted the cytoplasmic translocation of hnRNP K and to a lesser extent of hnRNP A2, meanwhile, JUNV infection induced an increase in hnRNP K cytoplasmic localization whereas hnRNP A2 remained mainly in the nucleus of infected cells. Both hnRNP K and hnRNP A2 were localized predominantly in the nucleus of JUNV persistently-infected cells even after superinfection with JUNV indicating that persistent infection does not alter nucleo-cytoplasmic transport of these hnRNPs. Total levels of hnRNP K expression were unaffected by DENV-2 or JUNV infection. In addition we determined, using small interfering RNAs, that hnRNP K knockout inhibits DENV-2 and JUNV multiplication. Our results indicate that DENV-2 and JUNV induce hnRNP K cytoplasmic translocation to favor viral multiplication.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2700157
Anti-HIST1H4A antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500309
Anti-Histone H4 (Acetyl-Lys12) antibody produced in rabbit, affinity isolated antibody
SAB4500310
Anti-Histone H4 (Acetyl-Lys16) antibody produced in rabbit, affinity isolated antibody
SAB4500307
Anti-Histone H4 (Acetyl-Lys5) antibody produced in rabbit, affinity isolated antibody
SAB4500308
Anti-Histone H4 (Acetyl-Lys8) antibody produced in rabbit, affinity isolated antibody
SAB4500306
Anti-Histone H4 antibody produced in rabbit, affinity isolated antibody
AV41035
Anti-HNRNPA2B1 antibody produced in rabbit, affinity isolated antibody
HPA001666
Anti-HNRNPA2B1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1405953
Anti-HNRNPK antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA007644 Anti-HNRNPK antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV40388
Anti-HNRPK (AB1) antibody produced in rabbit, affinity isolated antibody
AV40387
Anti-HNRPK (AB2) antibody produced in rabbit, IgG fraction of antiserum
WH0008365M1 Monoclonal Anti-HIST1H4H antibody produced in mouse, clone 6D4, purified immunoglobulin, buffered aqueous solution
SAB1403931
Monoclonal Anti-HNRNPA2B1 antibody produced in mouse, clone 3G11-6A7, purified immunoglobulin, buffered aqueous solution
WH0003181M1
Monoclonal Anti-HNRPA2B1 antibody produced in mouse, clone 1G12-6C5, purified immunoglobulin, buffered aqueous solution
NUC101
Nuclei Isolation Kit: Nuclei EZ Prep, sufficient for 25 nuclei preparations (~1-10×107 cells/preparation)
296988
Octane, anhydrous, ≥99%
C8H18
657042
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
C8H18