EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and chemistry
Detection of polycyclic aromatic hydrocarbons (PAHs) in raw menhaden fish oil using fluorescence spectroscopy: Method development.
PMID 25867932
Abstract
Raw menhaden fish oil was developed for biomonitoring polycyclic aromatic hydrocarbons (PAHs) using fluorescence spectroscopy. Menhaden (Genus Brevoortia) were collected in 2010 and/or 2011 from Delaware Bay, New Jersey, USA; James River, Virginia, USA; Vermillion Bay, Louisiana, USA (VBLA); and Barataria Bay, Louisiana, USA (BBLA). Barataria Bay, Louisiana received heavy oiling from the Deepwater Horizon oil spill. Method development included determining optimal wavelengths for PAH detection, fish oil matrix interferences, and influence of solvent concentration on extraction. Results showed that some fish oils contained high molecular weight PAH-like compounds in addition to other fluorescent compounds such as albumin and vitamin A and vitamin E. None of these naturally occurring compounds interfered with detection of high molecular weight PAHs. However, data suggested that the lipid component of fish oil was altering fluorescence spectra by supporting the formation of PAH excimers. For example, the most intense excitation wavelength for hydroxypyrene shifted from Ex285/Em430 to Ex340/Em430. Comparison of Deepwater Horizon crude oil and fish oil spectra indicated that some fish oils contained crude oil-like PAHs. Using wavelengths of Ex360/Em430, fish oil concentrations were calculated as 3.92 μg/g, 0.61 μg/g, and 0.14 μg/g for a Delaware Bay sample, BBLA 2011, and VBLA 2011, respectively. Overall, these results supported using menhaden fish oil to track PAH exposures spatially and temporally.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
361518
1-Hydroxypyrene, 98%
C16H10O
299847
2-Hydroxyfluorene, 98%
C13H10O
185507
2-Naphthol, 99%
C10H8O
130109
2-Naphthol, 98%
C10H8O
211281
9-Phenanthrol, technical grade
C14H10O
A2514
Albumin from goat, ≥96% (agarose gel electrophoresis)
A2639
Albumin from guinea pig serum, lyophilized powder, ≥96% (agarose gel electrophoresis)
A3060
Albumin from guinea pig serum, lyophilized powder, essentially fatty acid free
A5409 Albumin from hamster
A3139 Albumin from mouse serum, lyophilized powder, ≥96% (agarose gel electrophoresis)
A3559 Albumin from mouse serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
A1056
Albumin from mouse serum, essentially fatty acid free
A4414 Albumin from porcine serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
A2764 Albumin from porcine serum, lyophilized powder, 96-99% (agarose gel electrophoresis)
A1830
Albumin from porcine serum, lyophilized powder, ≥98% (agarose gel electrophoresis)
A0764
Albumin from rabbit serum, lyophilized powder, essentially globulin free, ~99% (agarose gel electrophoresis)
A0639
Albumin from rabbit serum, lyophilized powder, ≥96% (agarose gel electrophoresis)
A9438 Albumin from rabbit serum, essentially fatty acid-free
A6272
Albumin from rat serum, lyophilized powder, ≥96% (agarose gel electrophoresis)
A4538
Albumin from rat serum, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
A6414
Albumin from rat serum, lyophilized powder, essentially fatty acid free, essentially globulin free, ≥99% (agarose gel electrophoresis)
A2018 Albumin from rat serum, essentially fatty acid free
A3264
Albumin from sheep serum, lyophilized powder
A6289 Albumin from sheep serum, essentially fatty acid free
B2518
Albumin, Acetylated from bovine serum, lyophilized powder, suitable for (for molecular biology)
B8894
Albumin, Acetylated from bovine serum, protease free, for molecular biology
SCB-002
Benzo[b]fluoranthene, vial of 1 g, analytical standard
C20H12
275336
Benzo[b]fluoranthene, 98%
C20H12
05470
Bovine serum albumin, lyophilized powder, crystallized, ≥98.0% (GE)
A2153
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)