EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Rational Topological Design for Fluorescence Enhancement upon Aggregation of Distyrylfuran Derivatives.
PMID 25868005
Abstract
A series of 2,5-distyrylfuran derivatives bearing pentafluorophenyl- and cyanovinyl units have been synthesized for aggregation-induced emission (AIE). The effect of the type and extent of the supramolecular connections on the AIE of the furan derivatives were examined and correlated with their X-ray crystal structures. It was found that the simultaneous presence of cyano and perfluorophenyl units strongly enhances the fluorescence upon aggregation. Single-crystal X-ray diffraction analysis confirmed that CH⋅⋅⋅F, F⋅⋅⋅F, CH⋅⋅⋅nitrile, Ar⋅⋅⋅ArF (Ar=aryl, ArF =fluoroaryl), and nitrile⋅⋅⋅ArF intra- and intermolecular interactions drive the topology of the molecule and that solid-state supramolecular contacts favor AIE of the furan derivatives.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
146056
3-Nitrobenzyl alcohol, 98%
C7H7NO3