EMAIL THIS PAGE TO A FRIEND
International journal of food microbiology
Characterization of in vitro antifungal activities of small and American cranberry (Vaccinium oxycoccos L. and V. macrocarpon Aiton) and lingonberry (Vaccinium vitis-idaea L.) concentrates in sugar reduced fruit spreads.
PMID 25868124
Abstract
In this study, cranberry and lingonberry concentrates were added to commercial sugar-reduced fruit spreads (raspberry-Aloe vera, strawberry-guava, and strawberry-lime), and tested for their antifungal activities. Selected strains of the species Absidia glauca, Penicillium brevicompactum, Saccharomyces cerevisiae and Zygosaccharomyces bailii, as well as xerophilic environmental isolates of the genera Penicillium and Eurotium were used for challenge testing. Initially, varying concentrations of synthetic antifungal agents, such as sodium benzoate, potassium sorbate and butyl 4-hydroxybenzoate were tested against these fungi on wort agar containing 31% fructose at different pH values. Subsequently, the experiments were conducted in fruit spreads containing different concentrations of cranberry and lingonberry concentrates. The results of this study demonstrate that these concentrates were able to inhibit growth of visible colonies of xerophilic and non-xerophilic fungi. Cranberry and lingonberry concentrates are interesting candidates for natural preservation against fungal growth in sugar reduced fruit spreads.